Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Formvar Carbon Support Film on Hexagonal Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
FCF100H-NI-UC FCF400H-CU-25 FCF200H-AU-EC FCF200H-NI-TB FCF400H-NI-TC FCF100H-CU-UA FCF200H-AU-SB FCF300H-CU-TC FCF100H-AU-UC FCF400H-AU-50 FCF200H-AU-UA FCF200H-CU-EC FCF400H-NI-UC FCF200H-NI-TC FCF300H-AU-UC FCF300H-NI-UC FCF300H-AU-UB FCF200H-CU-SC FCF100H-AU-UB FCF100H-CU-TB FCF400H-NI-EC FCF300H-NI-TC FCF400H-AU-UB FCF100H-CU-TC FCF300H-NI-50 FCF400H-CU-EC FCF100H-CU-UC FCF400H-AU-UA FCF200H-NI-EB FCF300H-AU-EC FCF200H-NI-25 FCF100H-CU-EC FCF300H-CU-UB FCF100H-NI-UB FCF100H-AU-UA FCF400H-AU-UC FCF300H-CU-EB FCF300H-CU-UA FCF200H-NI-UB FCF400H-AU-SB FCF100H-NI-EA FCF300H-AU-SC FCF400H-NI-TA FCF200H-NI-TA FCF400H-NI-EA FCF200H-CU-TA FCF400H-NI-UA FCF400H-NI-TB FCF400H-NI-SB FCF300H-AU-50 FCF200H-CU-TB FCF200H-AU-SC FCF200H-AU-TA FCF200H-NI-UA FCF100H-AU-SB FCF100H-CU-EA FCF400H-CU-EA FCF400H-CU-SC FCF300H-NI-25 FCF100H-NI-UA FCF400H-CU-TC FCF300H-AU-SB FCF400H-CU-SB FCF300H-AU-TC FCF200H-NI-SB FCF400H-CU-UB FCF400H-CU-EB FCF100H-CU-EB FCF300H-AU-25 FCF200H-CU-SB FCF400H-NI-EB FCF100H-NI-EC FCF200H-AU-TC FCF200H-NI-SC FCF400H-AU-SC FCF200H-CU-UC FCF100H-AU-EA FCF100H-CU-TA FCF100H-NI-25 FCF300H-AU-UA FCF300H-NI-UB FCF200H-AU-UB FCF200H-NI-EA FCF100H-CU-50 FCF300H-AU-EB FCF200H-AU-50 FCF400H-CU-TB FCF200H-NI-50 FCF200H-AU-UC FCF400H-AU-25 FCF300H-CU-SB FCF100H-AU-EC FCF100H-NI-TB FCF100H-AU-TB FCF100H-AU-SC FCF300H-CU-TA FCF100H-AU-TA FCF400H-NI-SC FCF100H-CU-SB FCF200H-NI-EC FCF100H-CU-25 FCF400H-CU-UA FCF400H-CU-TA FCF200H-CU-EA FCF400H-AU-TA FCF400H-AU-TB FCF400H-AU-TC FCF300H-CU-TB FCF400H-NI-50 FCF300H-NI-EA FCF300H-CU-UC FCF400H-AU-EB FCF200H-AU-25 FCF300H-AU-TA FCF400H-AU-EC FCF300H-NI-SB FCF300H-NI-TA FCF300H-NI-EC FCF300H-NI-EB FCF100H-NI-50 FCF300H-AU-TB FCF300H-CU-EC FCF400H-CU-50 FCF100H-AU-50 FCF400H-AU-EA FCF300H-CU-SC FCF200H-CU-25 FCF100H-NI-EB FCF100H-CU-UB FCF200H-CU-EB FCF100H-CU-SC FCF400H-NI-25 FCF400H-NI-UB FCF200H-CU-UB FCF100H-NI-TA FCF300H-AU-EA FCF300H-CU-25 FCF200H-AU-EA FCF100H-NI-SB FCF100H-AU-EB FCF200H-AU-TB FCF100H-NI-TC FCF100H-NI-SC FCF100H-AU-TC FCF200H-CU-50 FCF100H-AU-25 FCF300H-NI-UA FCF300H-NI-SC FCF300H-CU-EA FCF200H-CU-TC FCF200H-AU-EB FCF300H-NI-TB FCF200H-NI-UC FCF200H-CU-UA FCF400H-CU-UC FCF300H-CU-50
103305-052PK 204.45 USD InStock
103305-052 103303-168 103305-054 103305-050 103303-282 103303-160 103303-284 103303-162 103303-286 103303-164 103303-288 103302-918 103303-166 103303-170 103304-522 103304-520 103303-158 103303-312 103304-518 103304-516 103302-928 103303-150 103302-926 103303-152 103303-154 103303-310 103303-156 103302-920 103302-924 103302-922 103304-998 103304-996 103303-026 103303-300 103303-028 103303-302 103303-304 103303-306 103303-260 103303-380 103303-262 103303-020 103303-382 103303-022 103303-264 103303-384 103303-266 103303-024 103303-386 102100-470 103302-854 103302-852 102100-466 103302-856 102100-468 103303-308 102100-502 102100-504 102100-506 102100-508 103303-378 103305-086 103303-014 103305-000 103305-088 103303-258 103303-016 103303-018 103305-084 103305-006 103305-008 103305-002 103303-010 103305-004 103303-376 103303-012 103304-694 102100-472 103304-692 102100-474 102100-510 103304-690 102100-476 102100-512 103304-698 103304-696 103305-132 103305-010 103303-008 103302-718 103305-134 103305-012 103305-136 103304-682 103304-680 103302-438 103304-688 102100-526 103304-686 102100-528 103304-684 103304-956 103302-722 103302-720 102100-530 103302-444 102100-532 102100-534 102100-536 103304-954 103304-678 102100-538 103304-952 103304-950 103302-442 103302-440 103303-344 103303-346 103304-948 103304-946 103303-182 103303-184 103303-186 103303-188 103303-342 103302-930 102100-540 103303-190 102100-542 103303-192 103302-932 102100-544 103304-944 102100-546 103304-942 102100-548 103304-940 103303-172 103302-708 103302-706 103303-174 103303-294 103303-176 103305-046 103303-450 103303-296 103305-048 103303-178 103303-298 103303-452 103302-942 103302-940 103302-704 103302-702 103302-944 103303-180
Formvar Carbon Support Film on Hexagonal Grids, Electron Microscopy Sciences
Graticules Reticles and Grids
The formvar coated grids are stabilized with an evaporated carbon film. This type of coating is excellent for specimen support, especially for ultra thin sections. All film is laid on the shiny side of the grid.
  • Formvar coatings thickness of standard (10 nm), ultra-thin (5 to 6 nm), thick (15 to 20 nm), or extra thick (25 to 50 nm)
  • Select copper, nickel, or gold grids in 100, 150, 200, 300 or 400 mesh
  • Carbon coatings thickness of 1 nm, 3 to 4 nm, and 20 to 30 nm
  • Optically checked followed by batch testing in the EM
Packaging: Packed in grid storage box.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty