Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Formvar Support Film on Square Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
FF100-CU-ET FF100-NI-ET FF200-AU-UL FF150-CU-50 FF300-AU-50 FF100-NI-UL FF400-CU-50 FF150-AU-TH FF300-NI-25 FF200-CU-50 FF150-AU-ET FF200-CU-ET FF150-NI-TH FF100-AU-TH FF100-AU-25 FF100-NI-TH FF150-AU-UL FF300-NI-UL FF400-AU-25 FF100-NI-25 FF300-CU-50 FF400-CU-ET FF100-NI-50 FF150-CU-25 FF300-CU-TH FF100-CU-50 FF300-AU-UL FF200-CU-UL FF200-AU-50 FF200-AU-25 FF400-NI-UL FF200-NI-TH FF300-AU-ET FF300-CU-ET FF150-AU-50 FF150-CU-TH FF400-NI-TH FF300-NI-ET FF400-AU-ET FF400-CU-25 FF400-AU-UL FF100-AU-UL FF150-NI-ET FF200-NI-50 FF400-NI-25 FF400-NI-ET FF400-CU-UL FF200-NI-ET FF150-AU-25 FF100-CU-TH FF400-AU-TH FF150-CU-UL FF300-CU-UL FF200-AU-ET FF300-AU-TH FF400-AU-50 FF400-CU-TH FF150-NI-50 FF300-CU-25 FF200-CU-25 FF200-AU-TH FF200-CU-TH FF200-NI-25 FF150-CU-ET FF100-CU-UL FF150-NI-25 FF100-CU-25 FF400-NI-50 FF300-AU-25 FF100-AU-ET FF150-NI-UL FF200-NI-UL FF300-NI-TH FF300-NI-50 FF100-AU-50
103303-488PK 158.5 USD InStock
103303-488 103303-202 100502-062 103303-204 100502-064 103303-324 103303-326 103303-328 100491-148 100491-146 100502-066 103303-200 103303-486 100502-068 100491-150 103302-798 103302-236 103302-796 100491-154 100491-152 103302-230 103302-196 103302-234 103302-794 103302-232 103302-198 100502-070 103303-392 100491-136 103303-196 103303-394 100491-178 100491-134 103303-198 103303-396 100491-138 100491-140 103302-204 103302-202 100491-144 100491-142 100491-180 103302-200 103303-224 103303-388 103303-226 103303-228 103302-180 100491-168 103303-064 103303-220 103303-066 103303-222 100491-172 100491-170 100491-176 100491-132 103303-390 100491-174 103302-174 103302-172 103302-178 103302-176 103303-490 100491-158 100491-156 103303-492 103303-330 103303-494 103303-332 100491-162 100491-160 100491-166 100491-164 103302-228
Formvar Support Film on Square Grids, Electron Microscopy Sciences
Graticules Reticles and Grids
A thin film of pure formvar resin coats the shiny side of a square grid.
  • Formvar coatings thickness of standard (10 nm), ultra-thin (5 to 6 nm), thick (15 to 20 nm), or extra thick (25 to 50 nm)
  • Select from copper, nickel, and gold grids in 100, 150, 200, 300, and 400 mesh sizes
  • Optically checked followed by batch testing in the EM
Film is laid on the shiny side of the grid.
Packaging: Packed in grid storage box.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty