Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Carbon Film on Square Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
CF400-NI  CF150-NI-ET CF400-NI-50 CF400-NI-UL CF200-NI-50 CF400-NI-TH CF300-CU-UL CF150-AU-UL CF300-NI-UL CF200-NI-TH CF200-NI-UL CF400-CU-UL CF150-AU-50 CF300-AU-50 CF150-CU-TH CF300-AU-ET CF200-CU-25 CF400-AU-UL CF200-AU-50 CF300-NI-ET CF200-CU-TH CF150-CU-UL CF300-CU-TH CF400-AU-ET CF200-AU-ET CF200-AU-TH CF200-CU-UL CF200-CU-ET CF300-NI-TH CF150-NI-UL CF150-AU-25 CF400-AU-TH CF300-NI-50 CF400-CU-ET CF200-AU-UL CF150-NI-25 CF150-CU-50 CF300-AU-TH CF150-CU-ET CF300-AU-UL CF200-NI-25 CF400-CU-TH CF150-CU-25 CF150-NI-50 CF200-CU-50 CF150-AU-ET CF300-CU-25 CF400-NI-ET CF300-AU-25 CF300-CU-50 CF400-CU-25 CF400-AU-50 CF400-AU-25 CF400-CU-50 CF150-NI-TH CF200-AU-25 CF200-NI-ET CF300-NI-25 CF300-CU-ET CF150-AU-TH
100500-850BX 135.96 USD InStock
100500-850 100500-852 100500-854 100489-732 100489-730 103305-530 103303-640 100500-856 100500-858 103303-634 103303-636 103305-986 100489-722 103305-984 100489-720 100489-728 100489-726 100502-756 103305-988 100502-758 100489-724 100503-154 103303-638 100500-846 100500-848 103303-620 103303-422 103305-990 103303-424 103303-500 103303-502 100489-712 103305-998 100489-710 103305-996 103305-994 103303-420 100502-760 103303-540 103305-992 100489-718 100489-716 100489-714 103303-894 103303-498 103303-534 103303-896 103303-536 103303-538 103303-614 103305-964 103305-962 103303-890 103303-496 103303-892 100489-708 103305-966 103303-616 103303-418 103303-618
Carbon Film on Square Grids, Electron Microscopy Sciences
Graticules Reticles and Grids
A thin film of pure carbon is deposited on the shiny side of the grid.
  • Select copper, nickel, or gold grids in 150, 200, 300, or 400 mesh
  • Optically checked followed by batch testing in the EM
Carbon coatings thickness of Standard: Approx. 5-6nm, Ultra-Thin (UL): 3-4nm, Thick (TH): 10nm, Extra Thick (ET): 20-30nm.
Ordering information: Available in Standard (5 - 6 nm), Ultra-Thin (3 - 4 nm), Thick (10 nm) and Extra Thick (20 - 30 nm) film thickness.
Packaging: Packed in grid storage box.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty