Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Carbon Film on Hexagonal Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
CF400H-CU-25 CF200H-AU-TH CF300H-NI-25 CF400H-NI-25 CF400H-NI-ET CF300H-CU-ET CF200H-NI-25 CF400H-AU-50 CF200H-NI-ET CF100H-NI-ET CF100H-NI-50 CF200H-NI-UL CF200H-AU-UL CF300H-AU-ET CF100H-AU-TH CF400H-NI-TH CF200H-CU-25 CF400H-CU-TH CF100H-AU-UL CF300H-CU-UL CF400H-NI-50 CF100H-CU-ET CF100H-NI-25 CF400H-CU-50 CF300H-AU-50 CF100H-AU-ET CF200H-CU-50 CF300H-CU-TH CF200H-CU-ET CF400H-AU-TH CF400H-AU-ET CF100H-AU-50 CF200H-CU-UL CF200H-CU-TH CF100H-CU-25 CF400H-CU-ET CF100H-AU-25 CF400H-AU-25 CF300H-NI-UL CF200H-AU-ET CF300H-NI-ET CF200H-NI-50 CF200H-AU-25 CF200H-NI-TH CF100H-CU-UL CF200H-AU-50 CF300H-NI-TH CF100H-NI-UL CF100H-CU-TH CF300H-AU-UL CF300H-CU-25 CF400H-AU-UL CF300H-AU-25 CF400H-CU-UL CF300H-CU-50 CF100H-CU-50 CF300H-AU-TH CF400H-NI-UL CF100H-NI-TH CF300H-NI-50
102099-974BX 94.96 USD InStock
102099-974 103303-520 102099-972 102099-970 103305-534 103305-536 103305-974 102099-978 103305-972 103305-532 102099-976 103305-970 102100-096 103305-538 102100-006 102100-008 103304-008 103303-630 103303-632 103303-514 103303-516 102099-980 103305-544 103305-546 103305-540 103305-542 102100-010 102100-098 102100-012 102100-014 102100-016 103303-518 103303-542 103303-544 103303-546 103303-626 103303-548 103303-460 102100-100 102100-102 102100-104 103303-628 102100-106 102100-108 103303-774 103303-454 103303-776 103303-456 103303-458 103303-770 103303-772 102100-110 103304-012 102100-112 103304-010 103305-968 102100-114 102100-116 102100-118 103304-014
Carbon Film on Hexagonal Grids, Electron Microscopy Sciences
Graticules Reticles and Grids
A thin film of pure carbon is deposited on the shiny side of the grid.
  • Carbon coatings thickness of standard (10 nm), ultra-thin (5 to 6 nm), thick (15 to 20 nm), or extra thick (25 to 50 nm)
  • Optically checked followed by batch testing in the EM
Packaging: Packed in grid storage box.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty