Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Formvar Carbon Support Film on Single Hole Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
FCFGA400-CU-UB FCFGA75-NI-EA FCFGA200-CU-EA FCFGA150-NI-SB FCFGA200-NI-SC FCFGA100-CU-EC FCFGA150-CU-EA FCFGA100-CU-SC FCFGA600-CU-EC FCFGA600-CU-EA FCFGA800-CU-EA FCFGA300-CU-25 FCFGA800-NI-SB FCFGA300-CU-50 FCFGA75-NI-EC FCFGA1000-CUSB FCFGA100-CU-TB FCFGA1000-NITB FCFGA600-NI-TB FCFGA400-CU-SB FCFGA400-NI-EA FCFGA600-NI-EC FCFGA400-NI-TC FCFGA600-CU-UB FCFGA300-NI-UC FCFGA75-CU-EA FCFGA400-CU-TC FCFGA300-NI-EB FCFGA400-CU-UC FCFGA75-CU-SC FCFGA200-NI-EA FCFGA200-CU-EB FCFGA1000-NITC FCFGA800-NI-UC FCFGA1500-NIEA FCFGA200-NI-SB FCFGA400-NI-TB FCFGA75-CU-UA FCFGA1500-CUTA FCFGA75-NI-SC FCFGA300-NI-TC FCFGA100-CU-SB FCFGA600-NI-EA FCFGA150-NI-50 FCFGA1500-NISB FCFGA300-NI-50 FCFGA75-NI-UC FCFGA1500-NIUB FCFGA150-CU-SC FCFGA150-CU-UB FCFGA600-CU-25 FCFGA100-NI-SC FCFGA75-CU-TB FCFGA1500-NISC FCFGA800-CU-SB FCFGA1500-NI-50 FCFGA1500-NITA FCFGA150-NI-UB FCFGA1500-CUTC FCFGA75-CU-UB FCFGA600-CU-SC FCFGA400-NI-TA FCFGA150-NI-SC FCFGA150-NI-EB FCFGA1500-CU-25 FCFGA600-NI-SC FCFGA100-NI-25 FCFGA800-NI-25 FCFGA1000-NITA FCFGA300-NI-TA FCFGA400-NI-UC FCFGA150-CU-SB FCFGA1000-CUUA FCFGA800-NI-50 FCFGA400-NI-50 FCFGA200-CU-UB FCFGA200-CU-25 FCFGA1500-CUUC FCFGA150-CU-UA FCFGA150-CU-EB FCFGA400-NI-EC FCFGA200-NI-EC FCFGA1500-CU-50 FCFGA800-CU-SC FCFGA1000-NI-50 FCFGA200-NI-UC FCFGA75-NI-EB FCFGA1500-CUEB FCFGA200-NI-50 FCFGA1000-CUEA FCFGA600-NI-50 FCFGA300-NI-25 FCFGA200-CU-50 FCFGA400-NI-UA FCFGA75-NI-50 FCFGA1000-CU-25 FCFGA150-NI-EA FCFGA100-NI-EA FCFGA200-NI-TC FCFGA800-NI-UB FCFGA400-CU-25 FCFGA75-CU-SB FCFGA100-CU-UC FCFGA1000-NIUB FCFGA1000-NIUA FCFGA75-CU-25 FCFGA1500-NITB FCFGA200-NI-EB FCFGA400-CU-EB FCFGA1000-NI-25 FCFGA1000-CUEB FCFGA1000-CUUB FCFGA300-NI-TB FCFGA100-NI-TB FCFGA1500-CUEA FCFGA400-CU-50 FCFGA300-CU-TA FCFGA800-NI-UA FCFGA75-CU-TC FCFGA600-CU-SB FCFGA600-NI-EB FCFGA150-CU-25 FCFGA300-CU-UC FCFGA200-CU-TB FCFGA150-NI-TB FCFGA600-CU-EB FCFGA100-NI-50 FCFGA75-NI-25 FCFGA300-NI-UB FCFGA1500-CUSC FCFGA100-CU-EB FCFGA100-CU-TC FCFGA150-CU-UC FCFGA800-CU-UB FCFGA75-NI-SB FCFGA600-NI-UC FCFGA400-CU-EC FCFGA200-CU-UC FCFGA300-NI-SB FCFGA200-CU-SC FCFGA75-CU-EB FCFGA200-NI-UB FCFGA200-NI-TB FCFGA800-NI-TB FCFGA75-CU-50 FCFGA300-CU-UB FCFGA800-CU-EB FCFGA800-CU-EC FCFGA400-CU-TA FCFGA200-CU-TC FCFGA75-NI-UB FCFGA100-NI-EC FCFGA150-CU-EC FCFGA1000-CUEC FCFGA600-NI-SB FCFGA300-CU-EB FCFGA200-CU-EC FCFGA400-CU-SC FCFGA1000-NIUC FCFGA600-CU-50 FCFGA600-CU-UA FCFGA400-NI-UB FCFGA400-CU-UA FCFGA800-NI-TC FCFGA100-NI-SB FCFGA100-CU-UB FCFGA200-NI-TA FCFGA100-CU-UA FCFGA400-NI-25 FCFGA200-CU-SB FCFGA1500-CUTB FCFGA150-NI-TA FCFGA400-NI-EB FCFGA800-CU-UC FCFGA400-CU-TB FCFGA75-NI-TA FCFGA300-NI-UA FCFGA300-CU-EA FCFGA150-NI-UA FCFGA75-CU-EC FCFGA1500-NIEC FCFGA800-CU-TB FCFGA100-NI-TC FCFGA800-CU-UA FCFGA600-CU-TB FCFGA1500-CUSB FCFGA1000-NISB FCFGA150-NI-EC FCFGA300-CU-SB FCFGA75-NI-TB FCFGA150-CU-TB FCFGA300-CU-SC FCFGA800-CU-TC FCFGA300-CU-TC FCFGA1000-CUTB FCFGA1500-CUUB FCFGA1000-NIEA FCFGA150-CU-TC FCFGA100-NI-EB FCFGA600-CU-UC FCFGA1000-CUUC FCFGA100-NI-TA FCFGA1500-NITC FCFGA300-CU-UA FCFGA100-NI-UB FCFGA1000-NIEC FCFGA800-CU-50 FCFGA150-NI-TC FCFGA300-NI-EA FCFGA1000-CUTC FCFGA800-NI-SC FCFGA75-NI-UA FCFGA200-NI-UA FCFGA100-NI-UC FCFGA75-CU-UC FCFGA1500-NI-25 FCFGA200-NI-25 FCFGA800-NI-EC FCFGA800-CU-TA FCFGA1500-NIUC FCFGA100-CU-TA FCFGA600-CU-TC FCFGA300-CU-TB FCFGA200-CU-UA FCFGA1000-CU-50 FCFGA600-NI-UA FCFGA200-CU-TA FCFGA1000-CUTA FCFGA300-NI-EC FCFGA1500-NIUA FCFGA400-CU-EA FCFGA800-NI-EA FCFGA300-CU-EC FCFGA1500-NIEB FCFGA150-CU-50 FCFGA150-NI-25 FCFGA1000-NISC FCFGA75-CU-TA FCFGA800-CU-25 FCFGA1500-CUEC FCFGA150-NI-UC FCFGA75-NI-TC FCFGA600-NI-UB FCFGA300-NI-SC FCFGA600-CU-TA FCFGA400-NI-SB FCFGA800-NI-EB FCFGA1000-NIEB FCFGA400-NI-SC FCFGA100-CU-EA FCFGA600-NI-TC FCFGA100-CU-50 FCFGA100-CU-25 FCFGA100-NI-UA FCFGA800-NI-TA FCFGA600-NI-TA FCFGA1000-CUSC FCFGA600-NI-25 FCFGA1500-CUUA FCFGA150-CU-TA
103303-960PK 181.04 USD InStock
103303-960 103303-720 103303-722 103303-724 103304-648 102100-600 102100-602 103303-726 103304-646 102100-604 103304-644 103303-728 102100-606 103304-642 102100-608 103304-640 103303-950 103303-710 103303-952 103303-712 103303-714 103304-638 103304-198 103304-196 103304-194 103304-192 102100-610 103303-958 103304-636 103303-716 102100-612 103304-634 103303-718 102100-614 103304-632 102100-616 103304-630 102100-618 103304-628 103303-700 103304-626 103303-702 103304-190 103304-188 103304-186 102100-620 103304-184 103304-182 102100-622 102100-624 103304-624 103303-704 102100-626 103303-706 103304-622 102100-628 103304-620 103303-708 103303-930 103304-618 103303-932 103303-934 103304-180 103304-176 103304-174 103304-172 103304-170 103303-936 103304-178 103303-762 103303-764 103303-766 103303-768 103304-208 103302-838 103303-760 103302-832 103304-440 103302-830 103302-836 103302-834 103304-448 103304-206 103304-204 103304-446 103304-444 103304-202 103304-442 103304-200 103303-752 103303-994 103303-754 103303-996 103303-998 103303-756 103303-758 103303-990 103303-992 103303-750 103304-672 103304-430 103304-670 103304-438 103304-436 103304-434 103304-432 103303-740 103303-742 103303-986 103303-744 103303-746 103304-428 103303-988 103304-660 103304-668 103303-748 103304-426 103304-424 103304-666 103304-422 103304-664 103304-662 103304-420 103303-730 103303-732 103303-734 103303-736 103304-418 103302-828 103302-826 103304-650 103304-098 103304-096 103302-824 103304-094 103303-738 103304-658 103304-656 103304-654 103304-652 103303-686 103303-688 103303-680 103303-682 103303-684 102100-570 103304-122 103304-000 102100-560 103304-120 102100-562 102100-564 102100-566 102100-568 103304-128 103304-126 103304-004 103304-124 103304-002 103303-674 103303-676 103303-678 103303-670 103303-672 102100-580 102100-572 103304-232 103304-110 103304-230 102100-574 103304-472 103304-470 102100-576 102100-578 103304-118 103304-116 103304-236 103304-114 103304-234 103304-112 103302-738 102100-590 102100-592 103304-100 103304-462 102100-582 103304-220 102100-584 103304-460 103302-736 102100-586 103302-734 102100-588 103304-108 103304-228 103304-106 103304-468 103304-226 103304-104 103304-466 103304-224 103304-102 103304-464 103304-222 103304-452 102100-594 103304-210 103302-744 102100-596 103302-742 103304-450 102100-598 103302-748 103302-746 103304-218 103304-216 103304-458 103304-214 103302-740 103304-456 103304-212 103304-454 103303-920 103303-922 103304-166 103304-164 103304-162 103304-160 103303-924 103303-926 103303-928 103304-168 103304-154 103304-152 103304-150 103303-914 103303-916 103303-918 103304-158 103304-156 103304-144 103304-142 103304-140 103304-148 103304-146 103303-696 103303-698 103303-690 103303-692 103303-694 102100-550 103304-132 102100-552 103304-130 102100-554 102100-556 102100-558 103304-138 103304-136 103304-134
Formvar Carbon Support Film on Single Hole Grids, Electron Microscopy Sciences
Graticules Reticles and Grids
The formvar coated grids are stabilized with an evaporated carbon film. This type of coating is excellent for specimen support, especially for ultra thin sections. All film is laid on the shiny side of the grid.
  • Formvar coatings thickness of standard (10 nm), ultra-thin (5 to 6 nm), thick (15 to 20 nm), or extra thick (25 to 50 nm)
  • Carbon coatings thickness of 1 nm, 3 to 4 nm, and 20 to 30 nm
  • Select from copper and nickel with 75, 100, 150, 200, 300, 400, 600, 800, 1000, or 1500 µm holes
  • Optically checked followed by batch testing in the EM
Packaging: Packed in grid storage box.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty