Would you like to log in again?
45° Pin Mount and 90° Profile, Combination Holder, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
75950-07-B 75950-07-A
103304-086EA 194.01 USD InStock
103304-086 103304-380
45° Pin Mount and 90° Profile, Combination Holder, Electron Microscopy Sciences
Specimen Mounting Supplies
For use in Hitachi S-800, S-4000, S-4100, S-4200, S-4300, S-4500, S-4700 and S-3600N SEMs.
  • Aluminum or brass with stainless steel allen set screws
  • Holds SEM pin mounts
The scanning electron microscope holder is ideal for cross sections up to 6.35 mm (1/4") thick.
Delivery information: Includes allen wrench.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty