Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Formvar Carbon Support Film on Slots, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

FCF205-NI-25 FCF2010-AU-TA FCF2010-AU-EB FCF205-CU-UA FCF2010-NI-TA FCF205-AU-TB FCF2010-CU-EB FCF2010-CU-SB FCF2010-CU-UC FCF205-NI-50 FCF2010-AU-SC FCF2010-AU-25 FCF2010-AU-EA FCF2010-AU-TC FCF2010-NI-50 FCF205-NI-UC FCF2010-CU-EA FCF2010-CU-TC FCF205-NI-SC FCF205-CU-EB FCF2010-CU-50 FCF2010-CU-25 FCF205-AU-SC FCF2010-NI-EB FCF2010-CU-TB FCF205-NI-UA FCF205-CU-50 FCF2010-NI-UA FCF2010-NI-UC FCF205-CU-SB FCF2010-AU-UA FCF2010-NI-EC FCF205-NI-TC FCF2010-AU-SB FCF205-CU-25 FCF205-AU-UA FCF205-AU-TC FCF205-NI-SB FCF205-NI-TB FCF2010-CU-UB FCF205-AU-EA FCF205-AU-50 FCF205-AU-TA FCF2010-NI-SB FCF2010-NI-TC FCF205-CU-UC FCF205-CU-TA FCF2010-NI-EA FCF205-AU-UB FCF205-CU-TB FCF2010-AU-UB FCF2010-AU-TB FCF205-CU-EA FCF205-NI-EA FCF2010-AU-UC FCF205-NI-EB FCF205-AU-EC FCF205-AU-EB FCF2010-CU-UA FCF205-AU-SB FCF2010-NI-25 FCF2010-AU-50 FCF205-NI-UB FCF2010-NI-SC FCF205-CU-EC FCF205-CU-UB FCF2010-CU-SC FCF2010-AU-EC FCF2010-NI-TB FCF205-NI-TA FCF2010-CU-EC FCF2010-CU-TA FCF205-AU-UC FCF205-AU-25 FCF205-CU-TC FCF2010-NI-UB FCF205-CU-SC FCF205-NI-EC
103304-808PK 195.65 USD InStock
103304-808 103304-806 103304-804 103304-528 100491-104 103305-214 100491-102 103305-216 100491-108 100491-106 103304-880 100491-110 102100-520 103305-218 102100-522 103304-526 103304-888 102100-524 103304-524 103304-886 103304-884 103304-882 103305-220 103305-028 103305-348 103305-222 103305-026 103304-870 103304-594 103304-592 100491-100 103304-590 103304-878 103304-876 103304-754 103304-874 103304-752 103304-872 103304-750 103305-350 103305-030 103305-352 103304-748 103304-868 103305-276 103305-278 103304-584 103304-582 103304-580 103304-746 103304-866 103304-864 103304-588 103304-586 103305-440 103304-818 103305-280 103304-816 103304-814 103304-616 103305-204 103305-206 103305-442 103305-202 103305-444 103304-892 103304-890 103304-812 103304-614 102100-514 102100-516 103304-612 103304-810 103305-090 103304-896 102100-518 103304-894 103304-530
Formvar Carbon Support Film on Slots, Electron Microscopy Sciences
Graticules Reticles and Grids
The formvar coated grids are stabilized with an evaporated carbon film.

  • Formvar coatings thickness of standard (10 nm), ultra-thin (5 to 6 nm), thick (15 to 20 nm), or extra thick (25 to 50 nm)
  • Carbon coatings thickness of 1 nm, 3 to 4 nm, and 20 to 30 nm
  • Optically checked followed by batch testing in the EM

This type of coating is excellent for specimen support, especially for ultra thin sections. All film is laid on the shiny side of the grid.

Packaging: Packed in grid storage box.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty