Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Carbon Film on Single Hole Grids, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

CFGA100-NI-TH CFGA150-NI-UL CFGA600-CU-TH CFGA300-NI-UL CFGA75-NI-UL CFGA800-CU-UL CFGA75-NI-25 CFGA1000-NI-50 CFGA600-NI-UL CFGA150-CU-UL CFGA400-CU-UL CFGA300-NI-TH CFGA600-NI-50 CFGA150-NI-50 CFGA100-CU-UL CFGA800-NI-TH CFGA400-NI-ET CFGA1000-CU-50 CFGA800-NI-50 CFGA100-NI-50 CFGA200-NI-25 CFGA200-NI-ET CFGA300-NI-ET CFGA800-CU-ET CFGA600-CU-50 CFGA300-NI-50 CFGA100-NI-UL CFGA150-CU-TH CFGA800-NI-ET CFGA1500-NI-25 CFGA200-NI-50 CFGA75-NI-ET CFGA200-NI-UL CFGA300-CU-TH CFGA200-CU-50 CFGA1000-CU-TH CFGA1500-CU-50 CFGA1000-NI-ET CFGA1000-CU-UL CFGA600-NI-ET CFGA300-CU-25 CFGA200-NI-TH CFGA200-CU-25 CFGA75-NI-50 CFGA400-CU-25 CFGA800-CU-50 CFGA300-CU-ET CFGA1500-CU-ET CFGA200-CU-UL CFGA300-CU-UL CFGA75-CU-25 CFGA400-NI-TH CFGA400-NI-UL CFGA1000-CU-ET CFGA1500-NI-ET CFGA150-CU-25 CFGA75-CU-ET CFGA800-NI-UL CFGA600-CU-ET CFGA800-NI-25 CFGA100-CU-TH CFGA150-CU-ET CFGA1000-NI-TH CFGA200-CU-ET CFGA400-CU-TH CFGA100-NI-ET CFGA1500-CU-25 CFGA150-CU-50 CFGA75-NI-TH CFGA800-CU-TH CFGA1000-CU-25 CFGA75-CU-50 CFGA100-CU-25 CFGA1000-NI-25 CFGA1000-NI-UL CFGA400-CU-50 CFGA300-CU-50 CFGA150-NI-25 CFGA100-CU-50 CFGA400-NI-25 CFGA100-CU-ET CFGA800-CU-25 CFGA150-NI-ET CFGA600-NI-TH CFGA100-NI-25 CFGA75-CU-UL CFGA1500-NI-UL CFGA1500-CU-TH CFGA400-NI-50 CFGA1500-NI-TH CFGA600-NI-25 CFGA1500-NI-50 CFGA200-CU-TH CFGA600-CU-UL CFGA400-CU-ET CFGA600-CU-25 CFGA75-CU-TH CFGA300-NI-25 CFGA150-NI-TH CFGA1500-CU-UL
103304-928PK 202.95 USD InStock
103304-928 103304-926 103304-408 103304-924 103304-406 102100-174 103304-484 102100-200 103304-482 102100-202 103304-480 102100-204 102100-206 103304-922 103304-404 103304-800 103304-920 102100-208 103304-402 103304-488 103304-400 103304-486 102100-180 102100-182 102100-184 102100-176 102100-178 102100-210 102100-212 102100-214 102100-216 102100-218 103304-478 102100-190 102100-192 102100-194 102100-196 102100-220 102100-186 102100-222 102100-188 102100-224 102100-226 102100-228 103304-860 103304-858 102100-240 102100-198 102100-230 102100-232 102100-234 102100-236 103304-856 102100-238 103304-854 103304-852 103304-850 103304-848 103304-846 102100-250 102100-242 102100-244 102100-246 102100-248 103304-844 103304-842 103304-034 103304-792 102100-252 103304-396 103304-032 103304-790 103304-394 103304-030 103304-798 103304-038 103304-796 103304-036 103304-794 103304-398 103304-782 103304-022 103304-780 103304-020 103304-788 103304-028 103304-786 103304-026 103304-784 103304-024 103304-938 103304-936 103304-934 103304-490 103304-778 103304-932 103304-776 103304-930 103304-412 103304-410
Carbon Film on Single Hole Grids, Electron Microscopy Sciences
Graticules Reticles and Grids
A thin film of pure carbon is deposited on the shiny side of the grid.

  • Carbon coatings thickness of standard (10 nm), ultra-thin (5 – 6 nm), thick (15 – 20 nm), or extra thick (25 – 50 nm)
  • Select from copper and nickel with 75, 100, 150, 200, 300, 400, 600, 800, 1000, or 1500 µm holes

Optically checked followed by batch testing in the electron microscope.

Packaging: Packed in grid storage box.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty