Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Formvar Carbon Support Film on Thin Bar Square Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
FCFT300-CU-25 FCFT200-CU-UB FCFT400-NI-EA FCFT200-CU-EB FCFT400-NI-EB FCFT200-AU-UA FCFT1000-NI-SB FCFT200-CU-UA FCFT200-AU-EB FCFT400-CU-SC FCFT1000-NI-TA FCFT200-AU-EC FCFT300-NI-EB FCFT300-NI-EA FCFT300-AU-SB FCFT1000-CU-EA FCFT300-CU-UC FCFT1000-CU-TA FCFT200-AU-TC FCFT300-CU-TB FCFT300-AU-TA FCFT200-AU-TB FCFT300-NI-TA FCFT300-CU-SB FCFT1000-AU-SC FCFT400-AU-25 FCFT1000-NI-UC FCFT200-NI-TA FCFT300-CU-EC FCFT400-CU-50 FCFT400-AU-50 FCFT300-AU-EC FCFT1000-AU-UA FCFT400-AU-UB FCFT300-NI-UC FCFT1000-AU-TA FCFT300-CU-EA FCFT300-NI-50 FCFT400-AU-SC FCFT400-NI-TC FCFT300-CU-UA FCFT200-CU-SC FCFT400-NI-EC FCFT1000-CU-EB FCFT400-NI-UB FCFT300-AU-UC FCFT1000-AU-UC FCFT1000-NI-EA FCFT1000-CU-SB FCFT200-CU-TB FCFT200-CU-EA FCFT1000-AU-EC FCFT1000-AU-TB FCFT400-AU-TC FCFT400-CU-UB FCFT1000-CU-TB FCFT300-AU-SC FCFT200-NI-50 FCFT400-AU-SB FCFT200-AU-SC FCFT300-AU-TC FCFT200-NI-UA FCFT400-NI-UA FCFT300-NI-SC FCFT300-AU-EB FCFT200-NI-EB FCFT200-NI-25 FCFT200-NI-EC FCFT200-CU-EC FCFT400-CU-25 FCFT1000-CU-EC FCFT300-AU-EA FCFT200-CU-TA FCFT400-NI-SB FCFT200-AU-UC FCFT200-AU-EA FCFT1000-AU-UB FCFT1000-AU-SB FCFT200-AU-UB FCFT200-CU-UC FCFT1000-AU-25 FCFT400-CU-EC FCFT1000-NI-TB FCFT1000-CU-TC FCFT300-AU-UA FCFT200-NI-UC FCFT300-NI-UA FCFT1000-CU-UC FCFT300-CU-SC FCFT1000-AU-TC FCFT400-AU-TB FCFT200-CU-25 FCFT200-NI-TB FCFT1000-AU-EA FCFT200-AU-SB FCFT300-CU-50 FCFT1000-NI-TC FCFT300-AU-TB FCFT300-NI-TC FCFT400-CU-EB FCFT400-AU-TA FCFT300-CU-EB FCFT400-NI-50 FCFT400-NI-TB FCFT400-NI-25 FCFT1000-NI-50 FCFT200-NI-UB FCFT300-CU-UB FCFT1000-NI-UB FCFT1000-AU-50 FCFT400-NI-TA FCFT300-AU-UB FCFT300-AU-25 FCFT200-AU-TA FCFT200-NI-EA FCFT1000-CU-UA FCFT400-NI-UC FCFT300-AU-50 FCFT300-NI-UB FCFT1000-CU-50 FCFT300-NI-SB FCFT1000-AU-EB FCFT400-CU-UC FCFT400-CU-EA FCFT200-NI-SB FCFT300-NI-EC FCFT400-CU-UA FCFT200-AU-25 FCFT300-NI-25 FCFT1000-CU-25 FCFT400-AU-UA FCFT400-CU-SB FCFT400-AU-EA FCFT400-NI-SC FCFT200-CU-50 FCFT1000-NI-EB FCFT200-NI-TC FCFT300-CU-TC FCFT200-CU-TC FCFT400-AU-EB FCFT1000-NI-UA FCFT1000-CU-UB FCFT400-CU-TA FCFT200-NI-SC FCFT400-AU-UC FCFT400-CU-TC FCFT1000-NI-EC FCFT200-CU-SB FCFT400-CU-TB FCFT200-AU-50 FCFT1000-CU-SC FCFT1000-NI-25 FCFT300-CU-TA FCFT400-AU-EC FCFT1000-NI-SC FCFT300-NI-TB
103305-330PK 207.46 USD InStock
103305-330 103305-732 103305-734 103305-730 102100-690 102100-682 102100-684 102100-686 102100-688 103305-228 102100-692 102100-694 102100-696 103305-628 102100-698 103304-512 103304-510 103305-230 103304-508 103304-506 103305-632 103305-236 103305-634 103305-238 103305-232 103305-234 103305-630 103305-636 103305-638 103304-504 103304-502 103304-500 103305-240 103305-242 103305-644 103305-248 103305-646 103305-244 103305-640 103305-246 103305-642 103304-298 103304-296 103305-648 103304-294 103306-042 103304-292 103306-040 103306-046 103306-044 103305-250 103305-770 103304-726 103304-290 103305-138 103305-776 103305-016 103305-778 103305-018 103305-772 103305-650 103305-774 103305-014 103304-562 103304-286 103304-560 103304-284 103304-724 103304-722 103304-720 103304-962 103304-960 103304-288 103305-140 103305-142 103304-718 103304-958 103304-716 103304-714 103305-148 103305-144 103305-146 103304-550 103305-308 103304-558 103304-712 103304-556 103304-710 103304-554 103304-312 103304-552 103304-310 103305-152 102100-708 103303-982 103304-708 103305-154 103303-984 103304-706 103304-704 103305-150 103304-308 103305-314 103305-316 103305-712 103305-156 103305-310 103305-312 103305-158 103303-980 103305-718 103305-838 103305-714 103305-318 103305-716 102100-666 103305-836 103304-702 102100-668 103304-548 103304-306 102100-700 103304-304 103304-700 102100-702 103304-302 102100-704 103304-300 102100-706 103305-162 103305-164 103303-978 103305-160 103305-324 103305-720 103305-326 103305-722 103305-320 103305-166 103305-840 102100-680 103305-322 103305-168 103304-496 103305-728 102100-670 102100-672 103305-724 103305-328 102100-674 102100-710 103305-726 102100-676 102100-712 102100-678 103304-498
Formvar Carbon Support Film on Thin Bar Square Grids, Electron Microscopy Sciences
Graticules Reticles and Grids
The formvar coated grids are stabilized with an evaporated carbon film. This type of coating is excellent for specimen support, especially for ultra thin sections. All film is laid on the shiny side of the grid.
  • Formvar coatings thickness of standard (10 nm), ultra-thin (5 to 6 nm), thick (15 to 20 nm), or extra thick (25 to 50 nm)
  • Carbon coatings thickness of 1 nm, 3 to 4 nm, and 20 to 30 nm
  • Select copper, nickel, or gold grids in 200, 300, 400, or 1000 mesh
  • Optically checked followed by batch testing in the EM
Packaging: Packed in grid storage box.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty