Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Formvar Carbon Support Film on Thin Bar Hexagonal Grids, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

FCFTH600-CU-EA FCFTH400-AU-50 FCFTH600-NI-EA FCFTH200-CU-EA FCFTH300-CU-SC FCFTH300-AU-UB FCFTH400-AU-EA FCFTH200-CU-TC FCFTH600-AU-EC FCFTH400-AU-EC FCFTH300-AU-EA FCFTH200-AU-EC FCFTH600-NI-TB FCFTH200-CU-EB FCFTH300-NI-50 FCFTH300-AU-TA FCFTH300-AU-25 FCFTH200-AU-EA FCFTH400-AU-SC FCFTH600-CU-25 FCFTH400-CU-25 FCFTH200-NI-EA FCFTH600-NI-TA FCFTH200-CU-TB FCFTH600-CU-TA FCFTH400-NI-SC FCFTH400-NI-EC FCFTH300-AU-EB FCFTH300-NI-EB FCFTH400-AU-25 FCFTH400-CU-UC FCFTH600-NI-UA FCFTH200-AU-UB FCFTH300-CU-EA FCFTH400-NI-UA FCFTH600-NI-50 FCFTH400-CU-EA FCFTH200-AU-EB FCFTH600-CU-UC FCFTH400-CU-EC FCFTH300-AU-SB FCFTH300-CU-UC FCFTH400-NI-TB FCFTH400-NI-EB FCFTH400-NI-50 FCFTH200-NI-TC FCFTH600-CU-EB FCFTH400-CU-50 FCFTH300-CU-EB FCFTH300-CU-TA FCFTH400-AU-UA FCFTH600-CU-TB FCFTH600-CU-UA FCFTH300-AU-TC FCFTH400-NI-EA FCFTH600-AU-25 FCFTH600-AU-SC FCFTH600-CU-TC FCFTH300-AU-50 FCFTH200-AU-UA FCFTH600-AU-EB FCFTH600-AU-UC FCFTH400-CU-TC FCFTH600-NI-SC FCFTH400-CU-UB FCFTH300-CU-TC FCFTH400-AU-TB FCFTH400-NI-SB FCFTH200-AU-UC FCFTH600-AU-SB FCFTH400-NI-TC FCFTH400-AU-TA FCFTH300-CU-SB FCFTH600-NI-EC FCFTH600-CU-EC FCFTH200-CU-SB FCFTH200-AU-SB FCFTH200-NI-UC FCFTH200-CU-UC FCFTH200-AU-25 FCFTH300-NI-SC FCFTH300-AU-UA FCFTH300-CU-25 FCFTH400-AU-UB FCFTH600-NI-25 FCFTH200-CU-SC FCFTH200-AU-SC FCFTH300-AU-EC FCFTH200-CU-UA FCFTH200-AU-50 FCFTH600-AU-50 FCFTH400-NI-UC FCFTH300-CU-TB FCFTH300-NI-UB FCFTH300-AU-UC FCFTH200-NI-UA FCFTH300-NI-TA FCFTH300-NI-TB FCFTH400-AU-SB FCFTH600-CU-50 FCFTH200-NI-25 FCFTH400-NI-UB FCFTH200-NI-TB FCFTH400-NI-25 FCFTH300-NI-SB FCFTH400-CU-SB FCFTH600-AU-EA FCFTH600-NI-UB FCFTH300-NI-EC FCFTH300-NI-25 FCFTH600-CU-SB FCFTH300-CU-50 FCFTH600-AU-TA FCFTH200-NI-SB FCFTH600-AU-TB FCFTH600-NI-TC FCFTH600-NI-SB FCFTH200-NI-EC FCFTH300-NI-UA FCFH400-AU-50 FCFTH300-NI-EA FCFTH600-CU-SC FCFTH200-NI-TA FCFTH200-CU-50 FCFTH200-NI-50 FCFTH200-AU-TA FCFTH300-CU-UB FCFTH200-NI-EB FCFTH200-NI-UB FCFTH600-AU-UB FCFTH600-AU-TC FCFTH300-CU-EC FCFTH200-NI-SC FCFTH400-CU-SC FCFTH400-NI-TA FCFTH400-CU-EB FCFTH400-CU-TB FCFTH600-CU-UB FCFTH200-CU-TA FCFTH300-AU-TB FCFTH300-CU-UA FCFTH400-AU-TC FCFTH200-CU-EC FCFTH300-AU-SC FCFTH200-AU-TC FCFH400-AU-25 FCFTH200-CU-25 FCFTH400-AU-UC FCFTH300-NI-UC FCFTH200-CU-UB FCFTH600-NI-UC FCFTH200-AU-TB FCFTH400-CU-UA FCFTH400-CU-TA FCFTH600-AU-UA FCFTH600-NI-EB FCFTH400-AU-EB FCFTH300-NI-TC
103305-174PK 198.43 USD InStock
103305-174 103305-176 103303-962 103305-170 103303-964 103305-172 103303-966 103305-336 103305-852 103305-338 103305-854 103305-332 103305-178 103305-334 103305-850 103306-094 103306-098 103305-736 103305-856 103306-096 103305-738 103305-858 102100-720 103303-968 102100-722 102100-724 102100-726 102100-728 103305-184 103305-340 103305-186 103305-180 103305-182 103305-742 103305-346 103305-744 103305-860 103305-188 103305-342 103305-862 103305-344 103305-740 102100-730 103305-746 102100-732 103305-748 102100-734 103304-514 102100-736 103305-190 102100-738 103305-196 103305-198 103305-192 103305-194 103305-750 103304-342 103304-340 102100-740 102100-742 102100-744 103304-744 102100-746 103304-742 102100-748 103304-740 103305-880 103304-738 103304-736 103305-886 103305-882 103305-884 103304-572 103304-330 102100-750 103304-570 102100-752 102100-630 102100-754 102100-632 102100-756 103304-338 103304-734 102100-758 103304-578 103304-732 103304-336 103304-610 103304-576 103304-730 103304-334 103306-048 103304-574 103304-332 103305-252 103304-728 103304-968 103305-654 103305-258 103305-656 103305-254 103305-256 103305-652 103306-050 103304-320 103305-938 102100-760 103306-054 103305-658 103305-936 103306-052 103304-966 103304-328 103304-326 103304-964 103304-568 103304-566 103304-324 103304-322 103304-564 103305-020 103305-262 103305-022 103305-660 103305-264 103305-260 103304-318 103305-666 103305-942 103305-940 103305-668 103305-662 103305-266 103305-024 103305-268 103305-664 103306-102 103304-316 103306-100 103304-314 103305-272 103305-670 103305-274 103305-270 103305-676 103305-678 103305-672 103305-674 103305-680 103305-682 103305-842 103305-844 103305-200 103305-846 103305-848 102100-714 102100-716 102100-718
Formvar Carbon Support Film on Thin Bar Hexagonal Grids, Electron Microscopy Sciences
Graticules Reticles and Grids
The formvar coated grids are stabilized with an evaporated carbon film. This type of coating is excellent for specimen support, especially for ultra-thin sections.

  • Formvar coatings thickness of standard (10 nm), ultra-thin (5 to 6 nm), thick (15 to 20 nm), or extra thick (25 to 50 nm)
  • Carbon coatings thickness of 1 nm, 3 to 4 nm, and 20 to 30 nm
  • Select copper, nickel, or gold grids in 200, 300, 400, or 600 mesh
  • Optically checked followed by batch testing in the EM

All films are laid on the shiny side of the grid.

Packaging: Packed in grid storage box.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty