Would you like to log in again?
Brain Punch Kit, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
57404 57399 57395 57405 57400 57391 57401 57397 57393 57403 57402
103305-074EA 245.96 USD InStock
103305-074 103305-470 103305-792 103305-472 103305-076 103305-072 103306-266 103305-080 103305-474 103305-078 103303-970
Brain Punch Kit, Electron Microscopy Sciences
Biopsy Punches
The Brain Punch Kit is a set of electropolished and sharpened punches for the Palkovit's punch technique.
Each punch has an electro polished sharp end for smooth cuts and minimal tissue adhesion. The smaller tubes are reinforced by an outer layer of concentric tubing except at the tip.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty