Would you like to log in again?
Formvar Film on Thin Bar Square Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
FFT1000-NI-UL FFT400-AU-25 FFT1000-NI-50 FFT400-CU-ET FFT300-AU-TH FFT400-NI-50 FFT1000-AU-UL FFT400-CU-TH FFT200-CU-50 FFT200-AU-UL FFT300-CU-UL FFT200-NI-50 FFT300-CU-50 FFT1000-AU-ET FFT200-NI-ET FFT1000-NI-25 FFT200-CU-UL FFT200-AU-ET FFT300-NI-ET FFT400-NI-ET FFT200-CU-TH FFT1000-CU-UL FFT400-NI-UL FFT1000-NI-ET FFT200-CU-ET FFT1000-CU-50 FFT1000-CU-25 FFT400-CU-UL FFT1000-CU-TH FFT300-CU-TH FFT400-CU-25 FFT300-AU-50 FFT200-AU-TH FFT300-AU-25 FFT400-NI-TH FFT1000-AU-25 FFT200-CU-25 FFT1000-CU-ET FFT200-NI-25 FFT400-AU-TH FFT300-AU-UL FFT300-NI-25 FFT300-NI-TH FFT400-NI-25 FFT300-NI-UL FFT200-AU-25 FFT400-AU-50 FFT200-NI-TH FFT1000-NI-TH FFT400-AU-UL FFT300-CU-ET FFT1000-AU-TH FFT400-CU-50 FFT300-CU-25 FFT1000-AU-50 FFT200-NI-UL FFT300-NI-50 FFT200-AU-50 FFT400-AU-ET FFT300-AU-ET
103305-130PK 198.43 USD InStock
103305-130 103305-618 103304-282 102101-008 102101-006 103304-280 102101-004 102101-002 102101-000 103303-830 103303-832 103303-834 103305-622 103305-588 103305-624 103305-584 103305-620 103305-586 103304-276 103304-990 102100-970 102100-972 102100-974 102101-016 102100-976 103303-836 102100-978 102101-014 102101-012 103304-994 102101-010 103304-278 103304-992 103305-590 103304-070 103304-066 102100-980 103304-064 102100-982 102100-984 103306-036 103304-988 102100-986 103306-034 102100-988 103306-038 103304-068 103303-972 103303-974 103303-976 103305-082 103305-126 103305-602 103305-128 103305-124 102100-990 102100-992 102100-994 102100-996 102100-998
Formvar Film on Thin Bar Square Grids, Electron Microscopy Sciences
Graticules Reticles and Grids
A thin film of pure formvar resin coats the shiny side of a Thin Bar Square grid. Coated grids are optically checked and batch tested in the electron microscope.
  • Select from copper, nickel, and gold grids in 200, 300, 400 or 1000 mesh sizes
  • Optically checked followed by batch testing in the EM
Formvar coatings thickness of standard (10 nm), ultra-thin (5 to 6 nm), thick (15 to 20 nm), or extra thick (25 to 50 nm).
Packaging: Packed in grid storage box.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty