Would you like to log in again?
Formvar Film on Gilder Finder Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
FF200F1-AU-25 FF200F2-CU-50 FF200F1-NI-25 FF200F2-CU-25 FF200F1-CU-25 FF200F1-AU-UL FF200F1-NI-50 FF200F1-CU-TH FF200F1-NI-UL FF200F2-CU-UL FF200F1-NI-TH FF200F2-AU-TH FF200F1-CU-ET FF200F2-AU-25 FF200F2-AU-50 FF200F1-AU-TH FF200F2-CU-ET FF200F1-AU-50 FF200F2-NI-25 FF200F2-NI-ET FF200F1-AU-ET FF200F2-AU-ET FF200F1-NI-ET FF200F2-AU-UL FF200F2-NI-UL FF200F2-CU-TH FF200F2-NI-50 FF200F1-CU-50 FF200F1-CU-UL FF200F2-NI-TH
103303-884PK 167.49 USD InStock
103303-884 103303-268 103303-886 103305-390 102100-790 102100-780 102100-782 102100-784 102100-786 103303-290 103303-826 102100-788 103303-828 103303-270 103303-372 103305-304 103303-550 103303-374 103303-552 103305-388 102100-792 103305-928 103305-926 102100-794 102100-774 102100-796 103305-306 103302-724 102100-776 102100-778
Formvar Film on Gilder Finder Grids, Electron Microscopy Sciences
Graticules Reticles and Grids
A thin film of pure formvar resin coats the shiny side of a Gilder Finder grid.
  • Formvar coatings thickness of standard (10 nm), ultra-thin (5 to 6 nm), thick (15 to 20 nm), or extra thick (25 to 50 nm)
  • Select from copper, nickel, and gold slot Finder grids
Coated grids are optically checked and batch tested in the electron microscope.
Packaging: Packed in grid storage box.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty