Would you like to log in again?
Gilder High Mesh Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
G1500HH-NI G1000HH-CU G1500HH-AU G2000HA-CU G2000HA-NI G2000HA-AU G1000HH-AU G1000HH-NI G1500HH-CU
103302-424PK 107.77 USD InStock
103302-424 103302-666 103302-664 103302-422 103305-958 103302-668 103302-426 103305-468 103305-466
Gilder High Mesh Grids, Electron Microscopy Sciences
Graticules Reticles and Grids
These 1000, 15000 and 2000 mesh grids for transmsission electron microscopy.
  • Copper, nickel and gold grids
  • Mesh diameter is 2 mm
Each grid is individually inspected.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty