Would you like to log in again?
Formvar Carbon Support Film on Gilder Finder Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
FCF200F2-CU-EB FCF200F1-AU-EC FCF200F1-NI-EA FCF200F1-CU-EB FCF200F2-CU-EA FCF200F2-AU-25 FCF200F2-CU-TB FCF200F2-NI-UB FCF200F2-NI-EC FCF200F1-CU-TB FCF200F1-NI-50 FCF200F2-AU-EC FCF200F2-CU-UC FCF200F1-CU-TC FCF200F1-AU-UC FCF200F1-CU-UB FCF200F1-NI-TA FCF200F2-AU-TC FCF200F1-AU-UB FCF200F1-AU-25 FCF200F2-CU-25 FCF200F2-NI-TB FCF200F2-AU-EA FCF200F2-NI-EB FCF200F1-AU-SC FCF200F2-NI-UC FCF200F1-NI-UB FCF200F2-AU-TB FCF200F1-CU-TA FCF200F2-NI-50 FCF200F1-CU-SC FCF200F1-NI-SB FCF200F2-AU-50 FCF200F1-CU-25 FCF200F1-AU-EB FCF200F1-AU-TB FCF200F2-AU-UA FCF200F1-AU-TC FCF200F1-AU-SB FCF200F1-CU-UA FCF200F2-CU-SC FCF200F2-NI-UA FCF200F2-NI-25 FCF200F2-AU-UC FCF200F2-CU-TA FCF200F1-NI-UA FCF200F2-NI-EA FCF200F1-CU-UC FCF200F1-NI-UC FCF200F1-CU-EC FCF200F2-NI-TA FCF200F2-NI-SC FCF200F2-CU-SB FCF200F2-CU-UB FCF200F2-CU-EC FCF200F1-CU-EA FCF200F2-AU-SB FCF200F1-AU-50 FCF200F2-NI-SB FCF200F2-AU-SC FCF200F1-CU-50 FCF200F1-NI-25 FCF200F2-CU-50 FCF200F1-NI-TC FCF200F2-CU-UA FCF200F2-AU-UB FCF200F1-AU-UA FCF200F1-NI-EC FCF200F2-CU-TC FCF200F1-AU-TA FCF200F1-NI-TB FCF200F1-NI-EB FCF200F2-AU-TA FCF200F1-AU-EA FCF200F1-NI-SC FCF200F1-CU-SB FCF200F2-AU-EB FCF200F2-NI-TC
103305-372PK 234.73 USD InStock
103305-372 103303-366 103305-374 103303-522 103303-524 103305-370 103303-406 103303-526 103303-360 103303-880 103305-376 102100-490 103303-362 103303-882 103305-378 103305-498 102100-492 103303-364 102100-484 103304-242 102100-486 103304-240 102100-488 103303-408 103303-528 103304-248 103304-246 103304-244 103303-356 103305-382 103303-872 103303-358 103303-874 103305-384 103303-876 103303-878 103305-380 103305-500 103303-350 103305-502 103305-982 103303-352 103305-386 103303-354 103305-980 103303-870 102100-494 102100-496 103305-504 102100-498 103303-864 103303-940 103303-866 103303-942 103303-348 103303-868 103303-944 103305-478 102100-500 103305-482 103303-532 103303-410 103305-484 103303-412 103303-414 103305-480 103303-416 103305-368 103305-486 102100-480 103303-530 103305-488 102100-482 103304-250 102100-478 103304-416 103304-414 103303-938
Formvar Carbon Support Film on Gilder Finder Grids, Electron Microscopy Sciences
Graticules Reticles and Grids
The formvar coated grids are stabilized with an evaporated carbon film. This type of coating is excellent for specimen support, especially for ultra thin sections. All film is laid on the shiny side of the grid.
  • Formvar coatings thickness of standard (10 nm), ultra-thin (5 to 6 nm), thick (15 to 20 nm), or extra thick (25 to 50 nm)
  • Carbon coatings thickness of 1 nm, 3 to 4 nm, and 20 to 30 nm
  • Select copper, nickel, or gold grids
  • Optically checked followed by batch testing in the EM
Packaging: Packed in grid storage box.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty