Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Citrate Buffer (10X), pH 6.0, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences
64142-07 64142-08
103303-062EA 133.23 USD InStock
103303-062 103305-698
Citrate Buffer (10X), pH 6.0, Electron Microscopy Sciences
Buffers Biological Buffers
This buffer is intended for heat-induced antigen retriever on formalin-fixed paraffin-embedded (FFPE) tissue sections prior to application of antibodies.

  • 1X buffer solution intended for heat-induced antigen retriever in IHC
  • Use of this buffer on FFPE tissue section improves accessibility of antibodies to tissue antigens
  • Please refer to primary antibody protocol
  • Boiling point: 100 °C
  • Density: 0.9989 g/cm³ (8.336 lbs/gal.)

In IHC most commonly used fixative like formalin mask tissue antigens (cellular, membrane and nuclear) by cross-linking process, this results in poor or no staining in IHC.

Preparation of working solution: dilute this 10X buffer as needed (e.g. 90 ml of deionized or distilled water+10 ml of this buffer) mix well, 1X buffer can be stored at 2 to 8 °C.

The use of this buffer on FFPE tissue section improves accessibility of antibodies to tissue antigens.

Caution: Do not freeze.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty