Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
HRP-Conjugate Diluent, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62716 62717 62718-15 62718
103307-080EA 1756.01 USD InStock
103307-080 103304-006 103305-870 103306-246
HRP-Conjugate Diluent, Electron Microscopy Sciences
Buffers Assay Buffers
Antibody-diluent buffer with additional component for stabilizing HRP-conjugate.
  • For preparing solution of HRP-conjugate used as detection reagent
  • Antibody-dilutent buffer with additional component for stabilizing your HRP-conjugate
  • Can further standardize the assay preparing ready-to-use conjuigate solutions
Specifically designed for preparing solution of your HRP-conjugate used as the detection reagent. Allows user to further standardize the assay preparing ready-to-use conjugate solutions in advance and store them in refrigerator without loss of activity.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty