Would you like to log in again?
EMS Holey Carbon Film, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
HC300-AU HC200-NI-UL HC200-AU HC200-AU-UL HC200-CU-150 HC200-CU-100 HC400-AU-25 HC300-CU-150 HC300-NI-100 HC300-NI HC300-AU-100 HC400-AU-UL HC300-CU-100 HC300-AU-UL HC300-AU-25 HC400-NI-150 HC200-NI-150 HC400-NI-25 HC300-CU-UL HC400-NI-100 HC200-NI HC400-AU-150 HC200-CU-UL HC200-AU-150 HC300-NI-UL HC300-CU HC400-CU-UL HC400-CU-150 HC300-AU-150 HC300-NI-150 HC200-AU-100 HC400-CU-100 HC200-AU-25 HC200-NI-100 HC400-CU-25 HC200-CU HC400-AU-100
103305-570PK 264.17 USD InStock
103305-570 103305-572 103305-578 76439-760 103305-574 103305-576 76439-778 100501-076 76439-810 100501-074 103306-010 76439-752 100501-072 76439-794 103306-012 100501-078 103305-580 103306-008 103305-582 103305-864 76439-770 103305-566 103305-866 103305-568 76439-768 76439-744 76439-786 103305-868 103306-086 102101-358 103306-084 102101-356 102101-354 103306-088 102101-352 76439-802 102101-350
EMS Holey Carbon Film, Electron Microscopy Sciences
Graticules, Reticles and Grids
A thin film of carbon with round holes is laid on the shiny side of the grid.
  • Coated grids are optically checked followed by batch testing in the EM
Available on copper, nickel, and gold 200, 300, and 400 mesh grids.
Ordering information: Molybdenum grids available on request.
Packaging: Packed in grid storage box.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty