Would you like to log in again?
Carbon Film on Thin Bar Hexagonal Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
CFTH600-AU-50 CFTH300-CU-UL CFTH200-CU-25 CFTH600-CU-UL CFTH200-CU-50 CFTH600-AU-ET CFTH300-AU-TH CFTH400-NI-ET CFTH300-NI-UL CFTH300-AU-25 CFTH400-NI-UL CFTH400-AU-50 CFTH200-NI-UL CFTH600-AU-25 CFTH300-NI-25 CFTH400-AU-UL CFTH200-CU-UL CFTH300-AU-UL CFTH200-AU-50 CFTH600-CU-TH CFTH600-CU-ET CFTH400-CU-ET CFTH400-NI-TH CFTH200-CU-ET CFTH200-NI-25 CFTH200-AU-TH CFTH300-AU-ET CFTH600-NI-ET CFTH300-NI-50 CFTH400-NI-50 CFTH400-CU-25 CFTH300-AU-50 CFTH600-NI-25 CFTH200-NI-TH CFTH600-CU-50 CFTH600-NI-UL CFTH300-CU-ET CFTH400-AU-25 CFTH400-CU-50 CFTH200-NI-ET CFTH400-CU-UL CFTH300-CU-50 CFTH400-NI-25 CFTH400-CU-TH CFTH200-NI-50 CFTH300-NI-TH CFTH200-AU-UL CFTH300-NI-ET CFTH600-NI-50 CFTH300-CU-TH CFTH300-CU-25 CFTH600-CU-25 CFTH600-AU-UL CFTH200-CU-TH CFTH600-NI-TH CFTH200-AU-25 CFTH400-AU-ET CFTH400-AU-TH CFTH600-AU-TH CFTH200-AU-ET
102100-370BX 174.6 USD InStock
102100-370 102100-372 102100-362 103306-372 102100-364 102100-366 103305-978 102100-368 103306-134 103306-138 103306-136 103304-914 103304-836 103304-912 102100-380 102100-374 102100-376 102100-378 102100-334 102100-336 102100-338 103304-834 103306-024 103304-910 103304-832 103306-026 103306-118 103305-552 103305-554 103305-598 102100-350 102100-340 102100-342 103306-076 102100-344 102100-346 102100-348 103306-112 103304-986 103306-078 103304-984 103306-116 103304-982 103306-114 103305-600 103305-522 103305-524 102100-360 103305-520 102100-352 103306-082 103306-360 102100-354 103306-080 103306-362 102100-356 103306-120 103305-526 102100-358 103306-364
Carbon Film on Thin Bar Hexagonal Grids, Electron Microscopy Sciences
Graticules Reticles and Grids
A thin film of pure carbon is deposited on the shiny side of the grid.
  • Select copper, nickel, or gold grids in 200, 300, 400, or 600 mesh
  • Optically checked followed by batch testing in the EM
Carbon coatings thickness of standard (10 nm), ultra-thin (5 to 6 nm), thick (15 to 20 nm), or extra thick (25 to 50 nm).
Packaging: Packed in grid storage box.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty