Would you like to log in again?
K-kit Specimen Holder for Liquid TEM, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
K7260-402 K7260-602 K7260-420 K7260-620
103306-152PK 1167.14 USD InStock
103306-152 103306-150 103306-154 103306-148
K-kit Specimen Holder for Liquid TEM, Electron Microscopy Sciences
Specimen Mounting Supplies
K-kits are enclosed specimen sample holders designed to facilitate convenient TEM observation of liquid samples, allowing nanoobjects, aggregates, and agglomerates (NOAAs) in liquid samples to be characterized.
  • Can be used with most TEM holder brands
  • Strong structural reliability under vacuum
  • Sealing glue compatible to many solvents
Vacuum compatible sealing of liquids in electron-transmitting thickness, K-kits are micro reaction chambers for countless experiments in materials, chemical, and biological research.
Ordering information: Tools are optional available in a Tool Set or ordered individually. Glues are also available.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty