Would you like to log in again?
JEOL SEM Mount, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
75706 75707 75705 75708
103306-182PK 838.82 USD InStock
103306-182 103306-174 103302-164 103301-980
JEOL SEM Mount, Electron Microscopy Sciences
Specimen Mounting Supplies
We offer a complete line of SEM specimen mounts.
  • For scanning electron microscopy
  • Standard lathe finish and economically priced
  • Made from ultra-pure aluminum
Head: 50 mm diameter
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty