Would you like to log in again?
Orbital Specimen Holder, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
75911 75912
103306-180EA 444.67 USD InStock
103306-180 103306-178
Orbital Specimen Holder, Electron Microscopy Sciences
Specimen Mounting Supplies
Unique orbital specimen holder for a spin and raise SEM images to a whole new level.
  • Rotate 360° around the axis perpendicular to the electron beam
  • Position the sample with turn of a screw
  • Flexibile and superior control
The pot is coated to reduce noise, producing a black background behind the subject so every detail stands out, allowing the capture of stunning, clear images.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty