Would you like to log in again?
EMS Glycerol Mounting Medium With DAPI and DABCO, Electron Microscopy Science
Supplier: Electron Microscopy Sciences
17989-60 17989-61
103306-192EA 474.81 USD InStock
103306-192 103302-256
EMS Glycerol Mounting Medium With DAPI and DABCO, Electron Microscopy Science
Stains Microscopy and staining reagents
EMS Glycerol with DAPI and DABCO™ is an aqueous mounting medium for preserving fluorescence of tissue and cell smears.
  • Fortified with DAPI a counter-stain for DNA
  • Ready to use with anti-fading agent 1, 4-Diazobicyclo-(2,2,2-octane (DABCO™)
  • Used in situ hybridization techniques or other methods where fluorescence of DNA staining is required
  • RNA is also stained with DAPI
EMS Glycerol with DAPI and DABCO™ is an aqueous mounting medium for preserving fluorescence of tissue and cell smears. This unique formula prevents rapid photobleaching of FITC, Texas Red, AMCA, Cy2, Cy3, Cy5, Alexa fluoro 488, Alexa fluoro 594, Green fluorescent protein (GFP), tetramethyly rhodamine, Redox. Fluorescence is retained during prolonged storage at 4°C in the dark.
Caution: Not recommended for phycoethyrin (PE), phycocyanin (PC) and allophycocyanin (APC)
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty