Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Numbered Gunshot Residue Sampler, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

76480-16
103306-270KT 755.46 USD InStock
103306-270
Numbered Gunshot Residue Sampler, Electron Microscopy Sciences
Specimen Mounting Supplies
SEM mounts with adhesive tab secures in glass vial to avoid contamination.

  • Certified aluminum SEM mount
  • Specify 100 consecutive numbers between 0 and 9999
  • Vials are 25 mm OD × 55 mm high

Numbered, certified aluminum mount with a double-coated adhesive carbon tab applied to secure the sample in a clean, glass vial 25 mm OD x 55 mm high (1 × 2.17"). Packaged in a box of 100. Numbers can be specified as 100 consecutive numbers between 0 and 9999 (greatest numbers). Please inform customer service about the numbering sequence when ordering this product.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty