Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
C-Flat™ Holey Carbon Grids for Cryo TEM, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
C-Flat™
CF441-100-AU CFT423-50-AU CFT-223C-10 CF313-50-AU CFT424-100 CF-224C-100 CF342-100-AU CF212-25 CF424-50-AU CF222C-100-AU CF4MH-50 CF-222C-10-AU CF413-50-AU CF4MH-100 CF3MH-50-AU CF313-10-AU CFT213-100 CF-222C-50 CF412-100-AU CF-222C-10 CF213-10-AU CFT442-100 CF212-50-AU CF224C-50-AU CF442-100 CF422-50-AU CF413-50 CFT342-100-AU CF4MH-100-AU CF213-50 CF313-50 CF422-100 CFT212-50 CF422-50 CFT422-100 CF242-50-AU CF312-50-AU CFT412-100 CF3MH-100-AU CFT-222C-100 CF223C-50-AU CF423-50-AU CF442-25 CFT213-10 CF-223C-10-AU CF2MH-100 CF342-50-AU CFT2MH-50 CFT413-100 CF242-50 CF-222C-100 CF222C-50-AU CF213-100-AU CF442-50 CFT-222C-10 CF2MH-100-AU CFT422-50 CF2MH-50 CFT21-100 CF422-100-AU CF4MH-25 CF424-100 CF423-100-AU CF212-50 CFT413-50 CF213-25 CFT412-50 CF212-100 CFT2MH-100 CF-223C-10 CF442-100-AU CFT4MH-50 CFT3MH-50-AU CF442-50-AU CF213-100 CF242-100-AU CF412-50 CFT442-50 CF313-10 CFT-224C-100 CFT424-50 CFT213-50 CFT423-100-AU CF424-100-AU CF-222C-25 CF212-100-AU CFT242-50 CFT4MH-100 CF412-25 CFT3MH-100-AU CF224C-100-AU CF313-100-AU CF242-100 CFT-224C-50 CF413-100 CFT313-10 CF412-50-AU CFT342-50-AU CF2MH-50-AU CF213-10 CF413-100-AU CFT-222C-50 CF223C-100-AU CF424-50 CF413-25 CF-224C-50 CF412-100 CF4MH-50-AU CFT212-100 CF312-100-AU CF213-50-AU CF422-25
76499-764PK 1841.91 USD InStock
76499-764 102100-170 76499-766 102100-172 102100-050 76499-760 102100-052 76499-762 102100-164 102100-166 76499-756 102100-168 76499-758 103306-948 103306-944 103306-306 103306-308 103306-946 76499-752 102100-060 76499-754 102100-062 76499-750 102100-056 102100-058 76499-748 103306-304 103306-958 103306-956 76499-742 102100-070 76499-744 102100-072 76499-740 76499-738 102100-066 102100-068 76499-734 76499-736 103306-310 103306-848 103307-074 103306-966 103306-968 76499-730 76499-698 76499-732 102100-084 76499-694 76499-696 76499-728 102100-076 76499-724 76499-726 103306-282 103306-962 103306-288 103306-964 103306-960 103306-856 103306-978 103306-858 76499-720 102100-090 76499-722 102100-094 102100-130 76499-716 102100-088 103306-294 76499-718 103306-296 76499-712 MSPP-CF313-50 103306-290 76499-714 103306-292 103306-972 102100-128 103306-298 103306-970 103306-850 103306-866 103306-988 76499-710 102100-140 76499-706 102100-134 76499-708 102100-136 76499-702 102100-138 76499-704 103306-862 103306-984 103306-986 103306-980 103306-860 103306-982 103306-878 76499-700 102100-150 103305-794 102100-144 102100-146 102100-148 103306-874 103306-876 103306-990 103306-872 102100-162 102100-154
C-Flat™ Holey Carbon Grids for Cryo TEM, Electron Microscopy Sciences
Graticules Reticles and Grids
C-flat™ is an ultra-flat, holey carbon-coated TEM support grid for transmission electron microscopy (TEM).
  • Manufactured without plastics so it is clean and without residue
  • Better particle dispersion and more uniform thickness
  • Use for single particle analysis, cryo electron tomography and automated TEM analysis
C-flat™ leads to better data sets. Made with patent pending technology, C-flat™ provides an ultra-flat surface that results in better particle dispersion and more uniform ice thickness. Patterning is done using deep-UV projection lithography, ensuring the most accurate and consistent hole shapes and sizes down to submicron features. The precise methods by which C-flat™ is manufactured eliminate artifacts such as excess carbon and edges around holes.
C-flat™ holey carbon grids provide the ideal specimen support to achieve high resolution data in cryo-TEM making them an ideal choice for single particle analysis, cryo electron tomography and automated TEM analysis.
Cryo-electron tomography (cryoET) and Single Particle Analysis (SPA): Numerous researchers have reported that the ultra-flat surface of C-flat™ leads to even ice thickness and uniform particle distribution within the hole areas. This optimal particle distribution results in superior data being collected as compared with other holey support films. 2 µm hole sizes are standard but custom hole sizes are available so C-flat™ can accommodate the common magnifications used for quantitative TEM analysis.
Automated TEM: C-flat™ provides a regular array of analysis sites compatible with automated data collection software such as Leginon. This compatibility, in combination with the more uniform ice thickness and particle distribution reported by numerous researchers, results in more high-quality target sites per grid.
Product Line: C-flat™ is a holey carbon film supported by a standard TEM grid. C-flat™ products are fully specified by 4 parameters: the hole diameter and pitch of the holey carbon film and the material type and mesh size of the TEM grid. The breadth of applications in cryoTEM necessitate a wide range of holey carbon film patterns. And now, with the recent expansion of the product line, a C-flat™ holey carbon film is available for almost any application. Whether 600 nm holes are needed for very high magnifications with ultra-high resolution cameras or large open areas are needed for larger specimens, C-flat™ is the perfect holey carbon grid.
Ordering information: C-flat™ is immediately available in several standard array patterns including hole diameters/hole spacings of 0.6/2, 1/1, 1/2, 1/4, 1.2/1.3, 2/1, 2/2, 2/4, 4/2, and a multihole pattern. C-flat™ is supported by your choice of a 200 mesh or 400 mesh copper TEM grid and sold in quantities of 25, 50, or 100. C-Flat™ is available in a thick option that doubles the carbon thickness from approximately 20 to 40 nm. Thick C-flat product numbers end in -T, catalog numbers contain "CFT". Available in quantities of 50 and 100 per pack.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty