Would you like to log in again?
Carbon Film on Thin Bar Square Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
CFT1000-AU-TH CFT1000-NI-50 CFT200-NI-UL CFT200-CU-UL CFT400-AU-25 CFT200-AU-ET CFT200-AU-UL CFT200-NI-TH CFT300-CU-TH CFT1000-NI-TH CFT300-AU-UL CFT200-AU-TH CFT400-NI-ET CFT200-CU-50 CFT400-CU-25 CFT400-CU-50 CFT200-CU-25 CFT400-NI-TH CFT400-AU-UL CFT1000-AU-UL CFT200-NI-ET CFT1000-AU-ET CFT400-CU-ET CFT300-NI-TH CFT1000-CU-UL CFT300-CU-ET CFT200-AU-25 CFT400-CU-TH CFT400-NI-50 CFT200-CU-ET CFT300-CU-UL CFT300-AU-25 CFT400-AU-ET CFT1000-NI-25 CFT200-NI-25 CFT1000-CU-ET CFT300-NI-UL CFT300-CU-25 CFT400-NI-UL CFT1000-NI-ET CFT300-NI-ET CFT1000-CU-TH CFT200-AU-50 CFT1000-AU-50 CFT1000-CU-25 CFT1000-AU-25 CFT400-NI-25 CFT300-AU-TH CFT400-CU-UL CFT300-NI-25 CFT200-NI-50 CFT300-NI-50 CFT300-AU-50 CFT1000-NI-UL CFT400-AU-50 CFT1000-CU-50 CFT300-AU-ET CFT300-CU-50 CFT400-AU-TH CFT200-CU-TH
103305-976PK 228.07 USD InStock
103305-976 102100-290 102100-292 102100-294 102100-286 103306-370 102100-288 102100-320 102100-322 103306-130 102100-324 102100-326 102100-328 103306-132 102100-330 102100-296 102100-332 102100-298 103306-020 103305-548 103306-068 103306-022 103304-830 103304-908 103305-594 103304-906 103305-550 103304-828 103304-904 103304-826 103305-512 103305-874 103305-514 103305-876 103305-596 103306-072 103304-980 103306-070 103305-516 103305-878 102100-300 103306-074 103305-518 102100-302 102100-304 103306-358 103306-110 102100-306 102100-308 103304-978 102100-310 102100-312 102100-314 103306-124 102100-316 103304-976 103306-368 102100-318 103306-128 103306-366
Carbon Film on Thin Bar Square Grids, Electron Microscopy Sciences
Graticules Reticles and Grids
A thin film of pure carbon is deposited on the shiny side of the grid.
  • Select copper, nickel, or gold grids in 200, 300, 4000, or 1000 mesh
  • Optically checked followed by batch testing in the EM
Carbon coatings thickness of standard (10 nm), ultra-thin (5 - 6 nm), thick (15 - 20 nm), or extra thick (25 - 50 nm).
Packaging: Packed in grid storage box.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty