Would you like to log in again?
Glycerol Fluoro Mount with Para Phenylenediamine, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
17989-71 17989-70
103306-376EA 342.08 USD InStock
103306-376 103302-166
Glycerol Fluoro Mount with Para Phenylenediamine, Electron Microscopy Sciences
Stains Microscopy and staining reagents
EMS Glycerol Fluorescence Mount with strong anti-fading agent, 1, 4-phenylenediamine (PPD) is an aqueous mounting medium for preserving fluorescence of tissue and cell smears.
  • Ready to use mounting medium with dark coffee color and this color does not interfere with Immunofluorescence
  • Refractive Index: 1.4563
The unique formula prevents rapid photobleaching of FITC, Texas Red, AMCA, Alexa fluoro 488, Alexa fluoro 594, tetramethyly rhodamine, and Redox. The fluorescence is retained during prolonged storage at 4 °C in the dark. Application: Immunofluorescence, confocal microscopy. Storage: 2-8°C is recommended, Protect from light, DO NOT FREEZE
Caution: Not suitable for immunofluorescence of Cy dyes, Phycoerythrin (R-PE), phyocyanin (PC), and allophycocyanin (APC). DO NOT FREEZE.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty