Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Immuno Mount with Para Phenylenediamine, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences
17989-81 17989-80
103306-378EA 316.04 USD InStock
103306-378 103302-168
Immuno Mount with Para Phenylenediamine, Electron Microscopy Sciences
Stains Microscopy and staining reagents
EMS Immuno Mount™ with strong anti-fading agent, 1, 4-phenylenediamine (PPD) is an aqueous mounting medium for preserving fluorescence of tissue and cell smears.

  • Ready to use mounting medium with dark coffee color and this color does not interfere with Immunofluorescence
  • Refractive Index in solution: 1 400 ± 0.002

The unique formula prevents rapid photobleaching of FITC, Texas Red, AMCA, Alexa fluoro 488, Alexa fluoro 594, tetramethyly rhodamine, and Redox. The fluorescence is retained during prolonged storage at 4 °C in the dark. Applications in immunofluorescence and confocal microscopy.
Storage: 2-8°C, DO NOT FREEZE.

Caution: Not suitable for immunofluorescence of Cy dyes, Phycoerythrin (R-PE), phyocyanin (PC), and allophycocyanin (APC)

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty