Would you like to log in again?
QUANTIFOIL® UltrAuFoil Gold Holey Carbon Films, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Q350AR1A Q250AR2A Q350AR13A
103306-426PK 1361.26 USD InStock
103306-426 103306-434 103306-424
QUANTIFOIL® UltrAuFoil Gold Holey Carbon Films, Electron Microscopy Sciences
Graticules Reticles and Grids
These UltrAuFoil gold holey carbon films will reduce the movement of frozen specimens during imaging and used in electron cryomicroscopy.
  • Gold grids
  • Hole sizes of 1.2 microns
  • 200 or 300 mesh grids
This improves image contrast and quality leading to better 3D reconstructions with less data. During imaging at cryo-temperatures, traditional carbon supports move, particularly at the beginning of irradiation. This movement blurs images and makes it difficult to determine the structures of small and challenging molecules.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty