Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
S18 Brightfield Stage Micrometer, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
68043-35 68043-30 68043-40
103306-196PK 477.67 USD InStock
103306-196 103306-846 103307-102
S18 Brightfield Stage Micrometer, Electron Microscopy Sciences
Calipers
Ideal if used with both metric and English measurements.
  • Dual Scales of 25 mm or 1 inch
  • Graduated Divisions
  • High Definition, chrome on glass image
S18 Brightfield Stage Micrometer: Graduated Metric and English Scales for the calibration of measurement instruments across a wide range of magnifications.
This flexible graduated scale stage micrometer is suitable for low magnification, stereo inspection microscopes to higher magnification compound microscopes.
The scales are centered on all glass slide 76 × 25 × 1.5 mm thick.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty