Would you like to log in again?
Diamond Trimming Tool, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
TT-20 TT-90 TTR-90 TTR-45 TTR-20 CTTR CTT-20R TT-45
103307-118EA 3289.63 USD InStock
103307-118 103307-100 103306-324 102681-508 103307-120 103305-784 103306-322 102681-506
Diamond Trimming Tool, Electron Microscopy Sciences
Knives
Diamond trimming tools are trimming tools for room or cryo temperature
  • Produces pyramidal sides with an inclined angle of 45° or 20°
  • Epoxy or acrylate resin embedded biological samples
  • Use at Room temperature for metals, polymersnon-embedded brittle samples
  • Cryo temperatures of sucrose infiltrated and frozen hydrated sample
The trimtool 90 aids in the FIB cutting of biological and technical sample blocks, trimming the 90° inclined blocks sides and well as the surface of the block. The trimtool series of trimming tools supersedes all the cryotrim and ultratrim lines. Resharpening is still available for these lines.
These are used for trimming biological and materials samples. Due to the extreme sharpness of these diamond blades, less mechanical damage is applied to the sample during trimming.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty