Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ultra Diamond Knife, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

24-ULDR 18-ULDE 20-ULJR 35-ULDR 40-UL 40-ULD 35-US 21-USE 15-ULR 21-US 40-ULDR 15-ULDR 30-ULD 15-USR 15-US 27-ULR 18-USR 20-USJ 40-ULR 30-ULDR 40-ULE 30-UL 27-USR 30-USJ 30-US 24-US 24-ULE 18-USE 35-USE 35-ULR 27-US 18-ULD 15-ULDE 35-ULE 21-ULR 40-USE 30-ULE 24-UL 35-ULD 30-USJR 20-ULJ 30-ULJ 18-ULR 18-UL 21-ULE 24-ULD 27-UL 24-ULDE 20-USJR 24-USE 24-USR 15-ULE 30-ULR 30-USR 40-US 21-ULD 15-ULD 30-ULDE 30-ULJR 18-ULDR 27-ULE 24-ULR 21-USR 21-ULDE 35-ULDE 21-UL 40-ULDE 35-UL 35-USR 15-USE 18-US 18-ULE 40-USR 15-UL 30-USE 21-ULDR
103307-184EA 6164.37 USD InStock
103307-184 103307-182 102680-562 102680-682 103307-142 102680-560 102680-680 103307-148 102680-834 103307-108 102680-558 102680-832 102680-556 102680-830 102680-752 102680-750 102680-838 102680-836 103307-130 103307-178 103307-134 103307-176 102680-746 103306-580 102680-744 102680-788 102680-786 102680-784 103306-588 102680-828 102680-748 103306-586 103307-082 103307-162 103307-166 102680-782 102680-780 102680-582 102680-580 103307-202 102680-614 102680-734 102680-778 102680-612 102680-578 102680-732 103307-206 102680-610 102680-654 102680-730 102680-576 102680-652 102680-530 102680-618 102680-616 102680-736 103307-152 103307-196 103307-194 102680-650 102680-690 103307-158 102680-526 102680-646 102680-524 102680-644 102680-566 102680-522 102680-686 102680-564 102680-520 102680-684 102680-608 102680-728 102680-528 102680-648
Ultra Diamond Knife, Electron Microscopy Sciences
Knives
These knives are available in 35, and 45 degree angles. The optimal thickness range for cutting with these knives is between 30 and 150 nm. They are available in sizes ranging from 1.5 to 4.0 mm as a standard. All of these knives are tested within a thickness range of 30 to 150 nm.

The 45° recognized as the knife for routine sectioning

The 35° diamond knives have been used for reduced compression, smoother section surfaces and better structural preservation. Also these knives have proved advantageous in Lowicryls, inhomogenous samples such as undecalcified bone, dental materials, sectioning of soft industrial samples such as metals and polymers, as well as for hard and brittle samples, for example semiconductors (Si, GaAs, etc), superconducting oxides, nanocrystalline ceramics.

The ultra 35° knife (in the triangular holder) with a cutting range of 100 to 2 µm is used for dry sectioning of epoxy or acrylic resin embedded biological samples, which need to be investigated by element analysis. The gliding of the sections on the dry knife surface is facilitated with the use of our static line II ionizer.

The ultra diamond knife is now available in Jumbo with 35° and 45° angles and in 2.0 and 3.0 mm sizes.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty