Would you like to log in again?
RtcB Ligase
Supplier: New England Biolabs
M0458S M0458S
103307-256EA 84 USD InStock
CA103307-256 103307-256
RtcB Ligase
Nucleic Acid Reagents Cloning Kits and Ligation
RtcB Ligase is expressed as a His-tagged fusion in E. coli.
  • This is an Enzyme for Innovation (EFI)
  • These enzymes have interesting properties and unique specificities
RtcB Ligase from E. coli joins single stranded RNA with a 3'-phosphate or 2',3'-cyclic phosphate to another RNA with a 5' -hydroxyl. Ligation requires both GTP and MnCl₂ and proceeds through a 3'-guanylate intermediate. For substrates with a 2',3'-cyclic phosphate end, hydrolysis to a 3'-phosphate precedes 3' end activation with GMP and ligation.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty