Would you like to log in again?
Luna® Universal qPCR Master Mix, New England Biolabs
Supplier: New England Biolabs
Luna®
M3003E M3003S M3003S M3003L M3003E M3003L M3003X M3003X
76196-394EA 1164 USD InStock
CA103307-260 76196-394 103307-260 103501-470 CA103501-470 103307-258 CA103307-258 CA76169-432
Luna® Universal qPCR Master Mix, New England Biolabs
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
Rapid, sensitive and precise dye-based qPCR detection and quantitation of target DNA and cDNA sequences.
The NEB Luna Universal qPCR Master Mix is an optimized 2X reaction mix for real-time qPCR detection and quantitation of target DNA sequences using the SYBR®/FAM channel of most real-time qPCR instruments
The master mix formulation is supplied at 2X concentration and contains all PCR components required for amplification and quantitation of DNA except primers and DNA template. Genomic DNA or cDNA of interest can be quantitated with Luna qPCR, and existing as well as commercial qPCR assay primer sequences can be used.
It contains Hot Start Taq DNA Polymerase and has been formulated with a unique passive reference dye that is compatible across a variety of instrument platforms (including those that require a high or low ROX reference signal). It also features dUTP for carryover prevention and a non-fluorescent, visible dye to monitor reaction setup. This dye does not spectrally overlap with fluorescent dyes used for qPCR and will not interfere with real-time detection.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty