Would you like to log in again?
Bacillus laterosporus BbsI-HF, Restriction Enzymes, New England Biolabs
Supplier: New England Biolabs
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
R3539S R3539L R3539S R3539L
103258-506EA 323 USD InStock
CA103307-268 CA103258-506 103258-506 103307-268
Bacillus laterosporus BbsI-HF, Restriction Enzymes, New England Biolabs
Nucleic Acid Reagents Restriction Enzymes
An E. coli strain that carries the cloned and modified BbsI gene from Bacillus laterosporus (C. Story) is used as source.
One unit is defined as the amount of enzyme required to digest 1 µg of λ DNA in 1 hour at 37 °C in a total reaction volume of 50 µl.
Delivery information: It is supplied with CutSmart® Buffer (10X) and Gel Loading Dye, Purple (6X)
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty