Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LMP7/PSMB8 Mouse Monoclonal Antibody [clone: 1B3]

Supplier: Novus Biologicals
H00005696-M01
103308-484EA 798.46 USD InStock
103308-484
Anti-LMP7/PSMB8 Mouse Monoclonal Antibody [clone: 1B3]
Antibodies
The LMP7 / PSMB8 Antibody (1B3) from Novus Biologicals is a mouse monoclonal antibody to LMP7 / PSMB8. This antibody reacts with human. The LMP7 / PSMB8 Antibody (1B3) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: LMP7/PSMB8
Clonality: Monoclonal
Clone: 1B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty