Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MMP8 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-1913R-CY5
10332-646EA 811.1 USD InStock
10332-646
Anti-MMP8 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Matrix Metalloproteinase 8 (MMP8) is also known as neutrophil collagenase and collagenase 2. MMP8 degrades fibrillar collagens types I, II, III, aggrecan, serpins and alpha 2 macroglobulin. All collagenases cleave fibrillar collagens at one specific site resulting in generation of N terminal three quarter and C terminal one quarter fragments, which then denature to gelatin at body temperature. The substrate specificity of collagenases is variable: MMP1 degrades type III collagen more efficiently than type I or type II collagen, whereas MMP8 is more potent in degrading type I collagen than type III or type II collagen. MMP13, in turn degrades type II collagen 6 fold more efficiently than type I and type II collagens and displays almost 50 fold stronger gelatinolytic activity than MMP1 and MMP8. MMP8 is very similar to MMP1, sharing 57 % amino acid identity. Most cell types do not produce MMP8. Until recently, it was thought that MMP8 was produced exclusively by neutrophils, but it has also been detected in other cell types including arthritic chondrocytes and gingival fibroblasts. The human MMP8 gene has the chromosomal location of 11q22.2-22.3. MMP8 is heavily glycosylated, and the zymogen has a mass of 85 Kd. The zymogen is quickly activated to the 64 Kd form, and this breaks down to a cascade of active forms.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MMP8
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty