Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-KLK9 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-1967R-A350
10334-142EA 811.1 USD InStock
10334-142
Anti-KLK9 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Kallikrein 9, also known as Kallikrein-Like 3 (KLK-L3), is a chymotrypsin-like serine proteinase. Kallikrein 9 was discovered as the locus for kallikreins on chromosome 19 was more fully mapped and found by similarity to the other tissue kallikreins. Kallikrein 9 has been found in the ovary, thymus, testis, prostate, skin, breast and neuronal tissues and is made by many cell lines in culture. Kallikrein 9 levels in breast cancer and uterine cancer patients have been reported to drop as the disease progresses, thus hK9 might be considered a favorable prognostic marker. Different splice variants of hK9 have been reported, although it is not yet known if they produce functional proteins. The full length Kallikrein 9 encodes for a 250 amino acid protein, with a predicted mass of 27.5 kDa and a pI of 7.53. The 234 amino acid form predicts a protein of 26 kDa with a pI of 9.76 and this quite basic pI might give the shorter form a very different function or localization. The shorter sequence also diverges before the catalytic serine residue, making it unlikely to be proteolytically active. Pre-pro-kallikrein 9 has the 17 amino acid signal sequence is removed before secretion, and the Pro-kallikrein 9 is activated to Kallikrein 9 by removal of the 5 amino acid propeptide domain.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: KLK9
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty