Would you like to log in again?
Screw Cap Micro Tubes, Double Packed, Sarstedt
Supplier: Sarstedt
72.730.311 72.687.772
103348-840PK 69.62 USD InStock
103348-840 103218-901
Screw Cap Micro Tubes, Double Packed, Sarstedt
Tubes Microtubes
These micro tubes are ideal for pure, safe and aseptic handling of materials in clean and sterile rooms, test kits or highly sensitive analyses.
  • Made of PP
  • Sterile double packaging
Each bag is sub-packaged into a second additional bag. Removing the outer bag leaves a complete and sterile bag packaging for convenient processing in clean rooms or sterile environments. Packaging option is ideal for users working under clean room conditions to fill precious reagents into tubes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty