Would you like to log in again?
NanoXact Silver Nanospheres, 0.02 mg/ml, Citrate Surface, nanoComposix
Supplier: nanoComposix
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
AGCN40-500M AGCN30-500M AGCN30-100M AGCN50-100M AGCN10-500M AGCN60-500M AGCN40-100M AGCN100-500M AGCN20-500M AGCN80-100M AGCN20-100M AGCN200-500M AGCN100-100M AGCN50-500M AGCN10-100M AGCN60-100M AGCN80-500M
103370-060EA 775.38 USD InStock
103370-060 103370-040 103370-042 103370-064 103370-030 103370-052 103370-066 103370-022 103370-054 103370-046 103370-024 103370-034 103370-048 103370-036 103370-058 103370-028 103369-672
NanoXact Silver Nanospheres, 0.02 mg/ml, Citrate Surface, nanoComposix
Beads Microcarriers
High quality silver nanoparticles. Provided with full COA for great value. Displaceable citrate surface.
  • High quality nanoparticles, precisely engineered and highly characterized
  • Unagglomerated and monodisperse
  • Standard colloidal concentration (0.02 mg/mL, 20 ppm, 0.185 mM, OD's of 0.8 to 2.7 [depending on size])
  • Citrate surface is stable and displaceable
Spherical silver nanoparticles, 0.02 mg/mL
Each batch is extensively characterized to ensure it meets the highest quality standards, providing reliable and reproducible results. Products are supplied with COA's that include batch-specific TEM, DLS, Zeta Potential, UV-Vis, and pH measurements.
Colloidal silver has applications in Diagnostics, Antimicrobial coatings, Electronics and Photovoltaics, SERS, Optics, Nanotoxicology, Plasmonics (SPR/Surface Plasmon Resonance), novel Color Enhancements and Pigments, Molecular Detection, Sensors, and Photothermal and Microwave-based Therapies.
Custom formulations and ISO13485/cGMP compliant materials can be provided on request.
Delivery information: Ships via FedEx Priority Overnight (domestic). Cold packaging included. Due to temperature-sensitive nature of these materials, they will only be shipped during the week in order to prevent products from sitting in an uncontrolled temperature environment over the weekend.
Caution: DO NOT FREEZE. Store at 2 to 8 °C and away from light when not in use to maximize shelf life. Nanomaterials can be sensitive to high and low pH conditions and high salt concentrations. Use caution when changing environmental conditions as particles may aggregate.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty