Would you like to log in again?
Ready-to-Assay™ NTR1 Neurotensin Receptor Frozen Cells, Eurofins Pharma Discovery Services
Supplier: Eurofins Pharma Discovery Services
HTS034RTA
103382-840PK 1240.85 USD InStock
103382-840
Ready-to-Assay™ NTR1 Neurotensin Receptor Frozen Cells, Eurofins Pharma Discovery Services
Assays Cellular Assays
Human NTR1 / NTS1 GPCR frozen cells for Calcium Flux FLIPR Assays.
  • Calcium Flux Assay
  • Host Chem-1
  • Adherent rat hematopoietic cell line
  • Expresses endogenous Gα15 protein
Neurotensin is a 13 amino acid peptide that induces hypothermia and mediates analgesia when administered centrally, and also mediates the behavioral effects of antipsychotic drugs in rodents (Kinkead and Nemeroff, 2004). In addition, several intestinal functions, such as contractility and inflammation, are mediated by neurotensin (Castagliuolo et al., 1999). The actions of neurotensin are mediated by two GPCRs, NTR1 (or NTS1) and NTR2 (or NTS2). NTR1 knockout mice are resistant to the hypothermic, analgesic and contractile effects of neurotensin, indicating that NTR1 is the primary mediator of these effects (Pettibone et al., 2002). Small molecule antagonists, including SR48692 and SR142948A, that are partially selective for NTR1 also interfere with the biological effects of neurotensin (Labbé-Jullié et al., 1998). Cloned human NTR1-expressing cell line is made in the Chem-1 host, which supports high levels of recombinant NTR1 expression on the cell surface and contains high levels of the promiscuous G protein Gα15 to couple the receptor to the calcium signaling pathway. Thus, the cell line is an ideal tool for screening for antagonists of interactions between NTR1 and neurotensin.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty