Would you like to log in again?
Ready-to-Assay™ 5-HT2C Serotonin Family Receptor Frozen Cells, Eurofins Pharma Discovery Services
Supplier: Eurofins Pharma Discovery Services
HTS132RTA
103382-988PK 1240.85 USD InStock
103382-988
Ready-to-Assay™ 5-HT2C Serotonin Family Receptor Frozen Cells, Eurofins Pharma Discovery Services
Assays Cellular Assays
Human 5-HT2C GPCR frozen cells for Calcium Flux FLIPR Assays.
  • Calcium Flux Assay
  • Host Chem-1
  • Adherent rat hematopoietic cell line
  • Expresses endogenous Gα15 protein
5-Hydroxytryptamine (5-HT, also commonly known as serotonin) is synthesized in enterochromaffin cells in the intestine and in serotonergic nerve terminals. In the periphery, 5-HT mediates gastrointestinal motility, platelet aggregation, and contraction of blood vessels. Many functions of the central nervous system are influenced by 5-HT, including sleep, motor activity, sensory perception, arousal and appetite. A family of 12 GPCRs and one ion channel mediate the biological effects of 5-HT (Hoyer et al., 1994). 5-HT2C, which couples to Gq in most cells to stimulate intracellular calcium, is prominently expressed in brain and appears to modulate depression, anxiety and appetite (Miller, 2005; Serretti et al., 2004; Wood, 2003). The mRNA encoding 5-HT2C undergoes selective RNA editing that changes 4 amino acids in the second intracellular loop; these changes result in alteration of efficiency of coupling to G proteins. Alterations in editing of 5-HT2C have been detected in victims of suicidal depression and in mice treated with the SSRI, fluoxetine (Tohda et al., 2006). Cloned human 5-HT2C -expressing cell line is made in the Chem-1 host, which supports high levels of recombinant 5-HT2C expression on the cell surface for functional detection via the calcium signaling pathway. Thus, the cell line is an ideal tool for screening for agonists, antagonists and modulators at 5-HT2C.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty