Would you like to log in again?
Transport Tubes, Self Standing and Graduated, Precision Medical Devices
Supplier: Precision Medical Devices
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
123303 123200
103388-962CS 173.95 USD InStock
103388-962 103388-960
Transport Tubes, Self Standing and Graduated, Precision Medical Devices
Tubes Test Tubes
Designed for the transport of urine, serum, or other biological materials.
  • Molded graduations
  • Leak resistant screw caps included
  • Self standing with a conical bottom
  • Made of a super strong polypropylene copolymer
Transport tubes can be used to transport urine, serum, or other biological material or specimens. Yellow caps are often used for the transport of urine or specimens, and the black caps are used to transport toxicology related specimens.
Yellow or black caps can be used to indicate urine or toxicology testing.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty