Would you like to log in again?
Deoxynivalenol ELISA Assay Kit, Eagle Biosciences
Supplier: Eagle Biosciences
BTDOEK-001
103388-984EA 523.08 USD InStock
103388-984
Deoxynivalenol ELISA Assay Kit, Eagle Biosciences
Assays ELISAs
Deoxynivalenol ELISA , allows to detect the Deoxynivalenol in food and feed crop (cereals).
  • Dynamic Range: 3.1-120ng/ml
  • Sensitivity: 12ppb
  • Incubation: 0.75 hours
Deoxynivalenol (DON), also known as Vomitoxin, is a type of mycotoxin (toxic mould fungus). Chemically it belongs to type B trichothecenes. The mycotoxin DON is formed as a poisonous metabolite from the Fusarium genus, i.e. F. culmorum and F. graminearum. DON enters into the human food chain via contamination of plant-based foods. It is abundant in grains such as wheat, barley, oats, rye, maize and processed grains such as malt, beer and bread. The main toxin production by Fusarium species happens in the field. Climate conditions have a big influence on contamination.The harvest and storage conditions play also an important role in DON contamination. DON is a potent inhibitor of protein synthesis and thus acts generally cause cell damage.The mycotoxin DON has been connected with incidents of mycotoxicoses in both humans and farm animals.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty