Would you like to log in again?
RBM3 ELISA Assay Kit, Eagle Biosciences
Supplier: Eagle Biosciences
BTRBM-001
103388-990EA 1268.46 USD InStock
103388-990
RBM3 ELISA Assay Kit, Eagle Biosciences
Assays ELISAs
RBM3 ELISA provides highly sensitive and specific quantitative of RNA-binding protein 3 in samples.
  • Dynamic Range: 31-2000 pg/ml
  • Sensitivity: 10 pg/ml
  • Incubation: 2.5 hours
The RNA-binding protein 3 (RBM3) is a member of the glycine-rich RNA-binding protein family. The exact role of the cold shock protein RBM3 is not yet clear [2, 6]. The role of mRNA-binding proteins can be attributed to a regulation of mRNA splicing, stability, transport and ultimately the translation of mRNA into proteins. RBM3 consists of 157 amino acids with a predicted mass of 17 kD (1, 2) and is one of the few proteins that are upregulated at low temperature [3-5]. It is known that RBM3 acts especially in cellular stress (eg. hypothermia) as mRNA chaperones and protein translation stabilizer [1, 4, 5]. Chip et al. were able to show that RBM3 has anti-apoptotic properties in neurons [7] and recent studies show that RBM3 mediates structural plasticity and protective effects of neurodegeneration (8). For these reasons, RBM3 may play an important role as biomarker for many neurodegenerative diseases and could be used to monitor the effectiveness of therapeutic hypothermia. Several studies report RBM3 as prognostic marker in cancer or in linking stress-regulated RNA splicing to tumorgenesis. Low nuclear expression of RBM3 has been found to be associated with poor prognosis in several cancer forms e.g. breast, ovarian, colorectal, prostate cancer and malignant melanoma (9-14).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty