Would you like to log in again?
TNF alpha Neutralization Rate/Adalimumab ELISA Assay Kit, Eagle Biosciences
Supplier: Eagle Biosciences
R222
103388-996EA 1628.08 USD InStock
103388-996
TNF alpha Neutralization Rate/Adalimumab ELISA Assay Kit, Eagle Biosciences
Assays ELISAs
The recoveryELISA RTA is for research use only and not to be used in diagnostic procedures.
  • Dynamic Range: Adalimumab: 0.7 – 30 µg/mL; TNF alpha in sample without Adalimumab: 4-400ng/ml;TNF alpha in sample with Adalimumab: 4-160ng/ml
  • Sensitivity: Adalimumab: 0.7 µg/mL; TNF alpha: 4ng/ml
  • Incubation: overnight
Tumour necrosis factor (TNFα) is a signalling substance of the immune system which can cause, among other things, the formation of inflammatory mediators in cells. In inflammatory diseases such as rheumatoid arthritis, there is overproduction of TNFα, which can be inhibited by using therapeutic antibodies.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty