Would you like to log in again?
Human Obestatin ELISA Assay Kit, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
YK231
103389-014EA 1464.62 USD InStock
103389-014
Human Obestatin ELISA Assay Kit, Eagle Biosciences, Inc.
Assays ELISAs
Human Obestatin ELISAdetermines obestatin in plasma and serum samples.
  • Dynamic Range: 0.231 - 25ng/mL
  • Sensitivity: 0.231 ng/mL
  • Incubation: 24 hours
Obestatin is a 23 amino acid residues peptide isolated from the rat stomach. The peptide shares the precursor with a food intake stimulating peptide, ghrelin, but possesses reducing effects on food intake, gut motility and body weight (1). With the use of an antiserum directed against the mouse/rat obestatin, obestatin immunoreactivity (irOBS) was detected in cells of the gastric mucosa and myenteric plexus and in Leydig cells of the testis in Sprague–Dawley rats. Double labeling of myenteric plexus with antisera against obestatin and choline acetyltransferase (ChAT) revealed that nearly all irOBS neurons were ChAT positive and vice versa (2). Obestatin (100nM) added to dissociated and cultured rat cerebral cortical neurons elevated cytosolic calcium concentrations [Ca+2]i in a population of cortical neurons (2). Intracerebroventricular admini- stration of obestatin inhibited water drinking in ad libitum fed and watered rats, and in food and water deprived animals. In addition, obestatin inhibited angiotensin II-induced water drinking in animals provided free access to water and food (3). Obestatin peptides had no effect on insulin sensitivity as revealed by hypoglycaemic response when co-administered with insulin, supporting a role of obestatin in regulating metabolism through changes of appetite, but indicating no direct actions on glucose homeostasis or insulin secretion (4). It is supposed that in rats the effects of obestatin on food intake may be secondary to an action of the peptide to inhibit water drinking (3). Recently, it is reported affording cardioprotection to ischemic- reperfused isolated rat heart, inhibiting apoptosis in culture of similarly stressed cardiomyocytes(5) and inhibiting dopamine release in rat hypothalamus(6). The obestatin concerning study for energy homeostasis and body weight regulation could be expected to have a large development in the future. The Eagle Biosciences Human Obestatin ELISA Assay kit developed by our laboratory can be used for direct determination of blood obestatin level’s variations and will be a useful tool for further development of obestatin research.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty