Would you like to log in again?
Urinary Cortisol ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
DCM018
103389-080EA 379.23 USD InStock
103389-080
Urinary Cortisol ELISA, Eagle Biosciences
Assays ELISAs
Urinary Cortisol ELISA is a colorimetric method to determine free Cortisol concentration in urine.
  • Dynamic Range: 10 - 500 ng/ml
  • Sensitivity: 2.95 ng/ml
  • Incubation: 1.5 hours
Cortisol is a steroid hormone released from the adrenal cortex in response to an hormone called ACTH (produced by the pituitary gland), it is involved in the response to stress; it increases blood pressure, blood sugar levels, may cause infertility in women, and suppresses the immune system. Cortisol acts through specific intracellular receptors and has effects in numerous physiologic systems, including immune function, glucose-counter regulation, vascular tone, substrate utilization and bone metabolism. Cortisol is excreted primarily in urine in an unbound (free) form. Cortisol is bound, in plasma, from corticosteroid-binding globulin (CBG, transcotin), with high affinity, and from albumin. Only free cortisol is available to most receptors. These normal endogenous functions are the basis for the physiological consequences of chronic stress - prolonged cortisol secretion causes muscle wastage, hyperglycaemia, and suppresses immune / inflammatory responses. The same consequences arise from long-term use of glucocorticoid drugs. 
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty