Would you like to log in again?
Androstenedione Saliva ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
DCM027
103389-096EA 379.23 USD InStock
103389-096
Androstenedione Saliva ELISA, Eagle Biosciences
Assays ELISAs
Androstenedione Saliva ELISA is to detect Androstenedione concentration in saliva.
  • Dynamic Range: 20 - 1000 pg/ml
  • Sensitivity: 0.5 pg/ml
  • Incubation: 1.5 hours
Androstenedione has relatively weak androgenic activity, estimated at ~ 20% of testosterone. Secretion and production rates also exceed those of testosterone in women in whom significant extra-adrenal conversion of androstenedione to testosterone occurs. In premenopausal women the adrenal glands and ovaries each produce about half of the total androstendione (about 3 mg/day). After menopause androstenedione production is about halved, primarily due to the reduction of steroid secreted by the ovary. Nevertheless, androstenedione is the principal steroid produced by the postmenopausal ovary. The high serum-saliva correlation for androstenedione suggests that individual differences in serum androstenedione levels may be accurately estimated using saliva as a non-invasive alternative specimen.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty