Would you like to log in again?
Anti- MPO (P-ANCA) ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
DCM092
103389-110EA 719.23 USD InStock
103389-110
Anti- MPO (P-ANCA) ELISA, Eagle Biosciences
Assays ELISAs
Anti MPO (p-ANCA) ELISA measures IgG class autoantibodies against myeloperoxidase in serum,plasma.
  • Dynamic Range: 10 - 160AU/mL
  • Sensitivity: 0.73 AU/mL
  • Incubation: 2 hours
PR3- ANCA is the classical autoAntigens in Wegener´s granulomatosis with a clinical specificity of more than 95%. c-ANCA is documented to be present in different diseases. Anti-MPO antibodies are highly specific for idiopathic and vasculitis associated crescentic glomerulonephritis and also for classic polyarteritis nodosa, Churg-Strauss syndrome and the polyangitis overlap syndrome without renal involvement.7-10 With respect to sensitivity, either MPO or PR-3 antibodies were found in 77 to 100% of patients with idiopathic and vasculitis associated crescentic glomerulonephritis. In WG, anti-MPO antibodies were detected only occasionally and generally in patients negative for PR-3 antibodies.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty